Skip to Content
Merck
All Photos(1)

Key Documents

M-018

Supelco

Morphine-3-β-D-glucuronide solution

100 μg/mL in methanol: water ( 50:50(v/v)), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27NO9
CAS Number:
Molecular Weight:
461.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol: water ( 50:50(v/v))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

O[C@@H](C=C1)[C@H]2[C@](C1[C@H]3C4)(CCN3C)C5=C4C=CC(O[C@H]6[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O6)=C5O2

InChI

1S/C23H27NO9/c1-24-7-6-23-10-3-4-12(25)20(23)32-18-13(5-2-9(14(18)23)8-11(10)24)31-22-17(28)15(26)16(27)19(33-22)21(29)30/h2-5,10-12,15-17,19-20,22,25-28H,6-8H2,1H3,(H,29,30)/t10?,11-,12+,15+,16+,17-,19+,20+,22-,23+/m1/s1

InChI key

WAEXKFONHRHFBZ-RURUVCMVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A certified solution standard of Morphine-3-ß-D-glucuronide at a concentration of 100 µg/mL for low volumes of sample testing. This certified solution standard is suitable for LC/MS or GC/MS applications including forensic analysis or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenjiro Nagaoka et al.
Drug metabolism and pharmacokinetics, 27(4), 388-397 (2012-01-14)
UDP-glucuronosyltransferases (UGTs) are glycoproteins in endoplasmic reticulum membranes. UGT2B7 is an important UGT isoenzyme expressed in human liver and glucuronidates various endogenous and exogenous substances. Although this enzyme has three potential N-glycosylation sites (asparagine at positions 67, 68 and 315)
Shawn McCann et al.
Journal of opioid management, 6(2), 87-94 (2010-05-21)
The 3-glucuronide metabolites of morphine and hydromorphone have been implicated as a causative factor for patients exhibiting myoclonus. The primary goal of this study was to determine plasma levels of morphine-3-glucuronide (M3G) or hydromorphone-3-glucuronide (H3G) in patients demonstrating myoclonus and
Yoshitaka Hasegawa et al.
The Journal of pharmacy and pharmacology, 62(3), 310-314 (2010-05-22)
The aim was to investigate the pharmacokinetics of morphine and its metabolite, morphine-3-glucuronide (M3G), in a rat model of streptozotocin (STZ)-induced diabetes. Morphine (15 mg/kg) was administered intravenously, and the concentrations of morphine and M3G in the plasma, urine and
Arief Nurrochmad et al.
Drug metabolism and pharmacokinetics, 25(3), 262-273 (2010-07-09)
The formation of morphine-3-glucuronide (M-3-G, pharmacologically inactive) and morphine-6-glucuronide (M-6-G, active metabolite) by liver microsomes from humans and rodents, including chimeric mice carrying human liver, was evaluated in the presence of fatty acyl-CoAs. Medium- to long-chain fatty acyl-CoAs, including oleoyl-CoAs
Chang-Shu Zhan et al.
Fa yi xue za zhi, 26(3), 188-191 (2010-08-17)
To propose a method for simultaneous determination of codeine(COD), 6-monoacetyl-morphine (6-MAM), morphine (MOR), morphine-3-glucuronide (M3G) and morphine-6-glucuronide (M6G) in human urine by ultra performance liquid chromatography with tandem mass spectrometry (UPLC-MS/MS). After precipitation of protein by acetonitrile, the urine samples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service