Skip to Content
Merck
All Photos(1)

Key Documents

M-012

Supelco

(±)-MDA solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-3,4-Methylenedioxyamphetamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

NC(C)CC1=CC=C(OCO2)C2=C1

InChI

1S/C10H13NO2/c1-7(11)4-8-2-3-9-10(5-8)13-6-12-9/h2-3,5,7H,4,6,11H2,1H3

InChI key

NGBBVGZWCFBOGO-UHFFFAOYSA-N

General description

A certified solution standard suitable to use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. MDA, or 3,4-methylenedioxyamphetamine, is an illicit recreational drug of the amphetamine and phenethylamine classes. Also known as tenamphetamine, MDA is abused for its psychedelic and hallucinogenic effects, similar to those of MDMA (Ecstasy).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han-yi Zhu et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 31(11), 1858-1862 (2011-12-01)
To investigate the effect of piperphentonamine hydrochloride (PPTA) on cognitive deficits induced by ischemia-reperfusion and explore the possible mechanisms. SD rats were randomly divided into sham-operated group, ischemia-reperfusion group (with saline injection), PPTA-treated groups (2.5, 5, 10 mg/kg) and edaravone-treated
M Barenys et al.
Reproductive toxicology (Elmsford, N.Y.), 34(1), 57-65 (2012-03-07)
MDMA consumers are young people of childbearing age. Consequently, developmental exposure to this drug is a potential public health concern. Several studies have addressed MDMA neurotoxicity in adults; however, knowledge of the effects of MDMA on developing embryos is limited.
Camilla Montesano et al.
Journal of mass spectrometry : JMS, 48(1), 49-59 (2013-01-11)
This study describes a method for the screening of methylenedioxyamphetamine- and piperazine-derived compounds in urine by liquid chromatography-tandem mass spectrometry. These substances, characterized by possessing common moieties, are screened using precursor ion and neutral loss scan mode and then quantified
Keiko Nakanishi et al.
Analytical and bioanalytical chemistry, 404(8), 2427-2435 (2012-09-27)
A liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) procedure was developed for the simultaneous determination of enantiomers of the prevalent designer drug 3,4-methylenedioxymethamphetamine (MDMA) and its phase I and phase II metabolites in urine with chiral derivatization. The analytes in
Raul López-Arnau et al.
British journal of pharmacology, 167(2), 407-420 (2012-04-19)
Here, we have compared the neurochemical profile of three new cathinones, butylone, mephedrone and methylone, in terms of their potential to inhibit plasmalemmal and vesicular monoamine transporters. Their interaction with 5-HT and dopamine receptors and their psychostimulant effect was also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service