Skip to Content
Merck

Skip To

E-086

3-epi-25-Hydroxyvitamin D3 solution

50 μg/mL in ethanol, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
12352205
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

50 μg/mL in ethanol

mp

-144.0 °C

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@H](O)CCC3=C

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23-,24-,25+,27-/m1/s1

InChI key

JWUBBDSIWDLEOM-UEGFJKGBSA-N

General description

A Certified Spiking Solution® of 3-epi-25-Hydroxyvitamin D3, the epi isomer of the prohormone 25-hydroxyvitamin D3. The presence of 3-epi-25-hydroxyvitamin D3 can affect accurate quantitation of 25-hydroxyvitamin D3 levels by most routine LC-MS/MS methods and to an unknown extent in immuno- and protein binding assays. This solution standard is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls and calibrators for LC/UV and LC-MS/MS assays; as system tuning/suitability standards; linearity verification; internal proficiency testing; and for OEM use in preparation of reagents, development of assays and instrument IQ/OQ.

Application

3-epi-25-Hydroxyvitamin D3 may be used as a certified reference material (CRM) for the determination of the analyte in biological fluids by different chromatography techniques.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
739936739928753548
application(s)

clinical testing

application(s)

detection

application(s)

detection

application(s)

detection

format

single component solution

format

-

format

-

format

-

form

liquid

form

solution

form

solution

form

liquid

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

grade

certified reference material

grade

-

grade

-

grade

-

storage temp.

−20°C, −70°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

57.2 °F - closed cup

flash_point_c

14.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters