Skip to Content
Merck
All Photos(1)

Key Documents

739936

Sigma-Aldrich

3-epi-25-Hydroxyvitamin D3 solution

100 μg/mL in ethanol, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Assay

98% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

mp

-144.0 °C

application(s)

detection

storage temp.

−20°C

SMILES string

C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@H](O)CCC3=C

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23-,24-,25+,27-/m1/s1

InChI key

JWUBBDSIWDLEOM-UEGFJKGBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3-epi-25-Hydroxyvitamin D3 abbreviated as 3-epi-25(OH)D3 is a metabolite of vitamin D3 produced in the liver. 3-epi-25(OH)D3 is used as a biomarker to determine the status of vitamin D in the body.[1] It can be also employed as an analytical standard for the quantification of 3-epi-25(OH)D3 molecules in human serum using the LC-MS/MS method.[2]

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Full-scan mass spectral evidence for 3-epi-25-hydroxyvitamin D3 in serum of infants and adults
Stepman HCM, et al.
Clinical Chemistry and Laboratory Medicine, 49(2), 253-256 (2011)
Overestimation of 25-hydroxyvitamin D3 by increased ionisation efficiency of 3-epi-25-hydroxyvitamin D3 in LC-MS/MS methods not separating both metabolites as determined by an LC-MS/MS method for separate quantification of 25-hydroxyvitamin D3, 3-epi-25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 in human serum
van den Ouweland JMW, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 967, 195-202 (2014)
Juan Le et al.
Journal of lipid research, 60(5), 1058-1064 (2019-03-25)
Vitamin D metabolite analysis possessed significant clinical value for the pediatric department. However, invasive venipuncture sampling and high blood consumption inflicted much suffering on patients. For alleviation, we carried out a LC-MS method for 25-hydroxyvitamin D quantification in only 3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service