Skip to Content
Merck
All Photos(1)

Key Documents

E-060

Supelco

17β-Estradiol solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

CC12CCC3C(CCc4cc(O)ccc34)C1CCC2O

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1

InChI key

VOXZDWNPVJITMN-ZBRFXRBCSA-N

Gene Information

human ... ESR1(2099)

General description

A Certified Spiking Solution® suitable for use as starting material in calibrators, controls, or linearity standards for a variety of LC/MS or GC/MS applications from endocrinology to clinical and diagnostic testing.

17β-Estradiol belongs to the group of endogenous endocrine disruptors that disturb thyroid activity and function. It is also a sex hormone prescribed under the trade names Vagifem®, Estring®, Evamist, Elestrin®, Estrace®, and Estrogel® as a treatment for hypoestrogenism.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:
  • Electrochemical determination of 17β-Estradiol in river water samples using a reduced graphene oxide and a metal complex porphyrin-based sensor
  • Development of a fluorescence resonance energy transfer (FRET) based aptasensor to determine 17β-Estradiol in environmental water, urine, and milk samples
  • Analysis of milk product samples to estimate 17β-Estradiol by high-performance liquid chromatography (HPLC) coupled with fluorescence detection following its solid phase extraction using graphene/multi-walled carbon nanotubes (MWCNTs)
  • Solid phase extraction of 17β-estradiol and 17α-ethinylestradiol from tap water samples using magnetic graphene oxide (GO)/γ-Fe2O3 nanoparticles for their simultaneous quantification by high-performance liquid chromatography (HPLC) combined with fluorescence detector
  • Multi-residue analysis of 17β-estradiol and 17α-ethinylestradiol in commercial pharmaceutical formulation and urine samples using hanging mercury drop electrode and screen printed carbon electrode

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Elestrin is a registered trademark of Azur Pharma
Estrace is a registered trademark of Warner Chilcott Company, LLC
Estring is a registered trademark of Pfizer Health AB
Estrogel is a registered trademark of Ascend Therapeutics, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vagifem is a registered trademark of Novo Nordisk A/S Corp.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Piet et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(26), 10828-10839 (2013-06-28)
Circulating gonadal steroid hormones are thought to modulate a wide range of brain functions. However, the effects of steroid fluctuations through the ovarian cycle on the intrinsic properties of neurons are not well understood. We examined here whether gonadal steroids
William Rosner et al.
The Journal of clinical endocrinology and metabolism, 98(4), 1376-1387 (2013-03-07)
The objective of the study was to evaluate the current state of clinical assays for estradiol in the context of their applications. The participants were appointed by the Council of The Endocrine Society and charged with attaining the objective using
Mark A Tarnopolsky
Medicine and science in sports and exercise, 40(4), 648-654 (2008-03-05)
Women oxidize more lipid and less carbohydrate and protein compared with men during endurance exercise. The increase in fat oxidation is associated with higher intramyocellular lipid content and use as well as greater adipocyte lipolysis. Glucose rates of appearance and
Aniruddha Das et al.
Archives of sexual behavior, 43(7), 1403-1415 (2014-03-07)
Using nationally representative data from the 2005-2006 U.S. National Social Life, Health, and Aging Project, this study queried relationship, sexual, and sex hormone patterns among married evangelical women and men aged 57-85, relative to those in other religions. Results suggested
Angelo Di Leo et al.
Journal of the National Cancer Institute, 106(1), djt337-djt337 (2013-12-10)
At the time of the initial analysis of overall survival (OS) for the Comparison of Faslodex in Recurrent or Metastatic Breast Cancer (CONFIRM) randomized, double-blind, phase III trial, approximately 50% of patients had died. A final analysis of OS was

Questions

  1. これはヒトのエストラジオールで間違いないでしょうか? また、これをマウスなどに投与した際、マウスのエストロゲン受容体に対しては効果は出ないのでしょうか?

    1 answer
    1. This product is intended for research use only as an analytical standard. The solvent used to prepare this standard is acetonitrile which is not suitable for use in tissue culture or animal models.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service