Skip to Content

E-061

17β-Estradiol-D5 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H19O2D5
CAS Number:
Molecular Weight:
277.41
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-835-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])[C@]4([2H])O)[C@@H]3CCc2c([2H])c1O

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i3D,7D2,10D,17D

InChI key

VOXZDWNPVJITMN-MOKSCAJFSA-N

General description

A stable-labeled internal standard for 17ß-Estradiol testing or isotope dilution methods by GC/MS or LC/MS for endocrinology, clinical chemistry or diagnostic purposes. Estradiol is a sex hormone prescribed under the trade names Vagifem®, Estring®, Evamist, Elestrin®, Estrace®, and Estrogel® as a treatment for hypoestrogenism.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Elestrin is a registered trademark of Azur Pharma
Estrace is a registered trademark of Warner Chilcott Company, LLC
Estring is a registered trademark of Pfizer Health AB
Estrogel is a registered trademark of Ascend Therapeutics, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vagifem is a registered trademark of Novo Nordisk A/S Corp.


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library