Skip to Content
Merck

Skip To

A-072

Atenolol solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H22N2O3
CAS Number:
Molecular Weight:
266.34
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-835-2
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

CC(C)NCC(O)COc1ccc(CC(N)=O)cc1

InChI

1S/C14H22N2O3/c1-10(2)16-8-12(17)9-19-13-5-3-11(4-6-13)7-14(15)18/h3-6,10,12,16-17H,7-9H2,1-2H3,(H2,15,18)

InChI key

METKIMKYRPQLGS-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

General description

Atenolol, commonly sold under the trade name Tenormin®, is a beta blocker used to treat a number of cardiac conditions from hypertension to angina. This Snap-N-Spike® Reference Solution is suitable for use in atenolol methods by LC/MS or GC/MS for applications such as clinical toxicology, forensic analysis, or pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Tenormin is a registered trademark of Astrazeneca UK

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1909A13400001044403
description

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

European Pharmacopoeia (EP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

application(s)

clinical testing

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

form

liquid

form

powder

form

-

form

-

format

single component solution

format

-

format

neat

format

neat

Quality Level

300

Quality Level

300

Quality Level

-

Quality Level

-

grade

certified reference material

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard


Still not finding the right product?

Explore all of our products under Atenolol solution


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters