Skip to Content
Merck
All Photos(1)

Documents

A1340000

Atenolol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide, 4-[2′-Hydroxy-3′-(isopropylamino)propoxy]phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O3
CAS Number:
Molecular Weight:
266.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atenolol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)NCC(O)COc1ccc(CC(N)=O)cc1

InChI

1S/C14H22N2O3/c1-10(2)16-8-12(17)9-19-13-5-3-11(4-6-13)7-14(15)18/h3-6,10,12,16-17H,7-9H2,1-2H3,(H2,15,18)

InChI key

METKIMKYRPQLGS-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atenolol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atenolol: pharmacokinetic/dynamic aspects of comparative developmental toxicity.
Sonia A Tabacova et al.
Reproductive toxicology (Elmsford, N.Y.), 16(1), 1-7 (2002-04-06)
Ronald V Lacro et al.
The New England journal of medicine, 371(22), 2061-2071 (2014-11-19)
Aortic-root dissection is the leading cause of death in Marfan's syndrome. Studies suggest that with regard to slowing aortic-root enlargement, losartan may be more effective than beta-blockers, the current standard therapy in most centers. We conducted a randomized trial comparing
M McGuiness et al.
Journal of the American Academy of Dermatology, 37(2 Pt 2), 298-299 (1997-08-01)
Atenolol is a beta-blocker commonly used for treating hypertension. It can induce various kinds of adverse side effects, including psoriasiform skin eruptions, skin necrosis, vasculitis, and (rarely) drug-induced connective tissue disease. We encountered a patient receiving atenolol for his hypertension
A N Wadworth et al.
Drugs, 42(3), 468-510 (1991-09-01)
Atenolol is a selective beta 1-adrenoceptor antagonist with a duration of activity of at least 24 hours. The scope of therapeutic use of the drug has been expanded and become better defined since it was first reviewed in the Journal
Laura M Kuyper et al.
The Canadian journal of cardiology, 30(5 Suppl), S47-S53 (2014-04-23)
Previous reviews have shown that β-blocker use for the treatment of hypertension without compelling indications was associated with increased risk of stroke in the elderly. It remains unclear whether this increased risk was driven by the type of β-blocker. We

Articles

TLC plates, with silica gel, gypsum as binder, International Pharmacopoeia compliant, for identification of Atenolol and Chlorthalidone in tablets.

TLC plates, with silica gel, gypsum as binder, International Pharmacopoeia compliant, for identification of Atenolol and Chlorthalidone in tablets.

TLC plates, with silica gel, gypsum as binder, International Pharmacopoeia compliant, for identification of Atenolol and Chlorthalidone in tablets.

TLC plates, with silica gel, gypsum as binder, International Pharmacopoeia compliant, for identification of Atenolol and Chlorthalidone in tablets.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service