Skip to Content
Merck
All Photos(1)

Key Documents

LM2312

Avanti

Sphingomyelin (d18:1/12:0)

Avanti Research - A Croda Brand LM2312, ethanol solution

Synonym(s):

N-(dodecanoyl)-sphing-4-enine-1-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H71N2O6P
CAS Number:
Molecular Weight:
646.92
UNSPSC Code:
12352211
NACRES:
NA.25

form

ethanol solution

packaging

pkg of 1 × 1 mL (LM2312-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM2312

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C35H71N2O6P/c1-6-8-10-12-14-16-17-18-19-21-22-24-26-28-34(38)33(32-43-44(40,41)42-31-30-37(3,4)5)36-35(39)29-27-25-23-20-15-13-11-9-7-2/h26,28,33-34,38H,6-25,27,29-32H2,1-5H3,(H-,36,39,40,41)/b28-26+/t33-,34+/m0/s1

InChI key

HZCLJRFPXMKWHR-FEBLJDHQSA-N

General description

Sphingomyelin is one of the bioactive lipids found in plasma membrane.

Application

Sphingomyelin (d18:1/12:0) or N-(dodecanoyl)-sphing-4-enine-1-phosphocholine has been used as a lipid standard for phospholipid quantification by mass spectroscopy (MS) analysis.

Biochem/physiol Actions

Sphingomyelin (d18:1/12:0) or N-(dodecanoyl)-sphing-4-enine-1-phosphocholine serves as a substrate for sphingomyelinase.

Packaging

2 mL Amber Glass Sealed Ampule (LM2312-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Björkhem et al.
Journal of lipid research, 42(3), 366-371 (2001-03-20)
Infants with the cholesterol synthesis defect Smith- Lemli-Opitz syndrome (SLO) have reduced activity of the enzyme 7-dehydrocholesterol-7-reductase and accumulate 7-dehydrocholesterol, with the highest concentration in the brain. As a result of the generally reduced content of cholesterol, plasma levels of
Ngoc Vu et al.
Analytical and bioanalytical chemistry, 411(23), 5937-5949 (2019-07-08)
Dysregulated lipid species are linked to various disease pathologies and implicated as potential biomarkers for type 1 diabetes (T1D). However, it is challenging to comprehensively profile the blood specimen lipidome with full structural details of every lipid molecule. The commonly
Ali Assi et al.
Analytical and bioanalytical chemistry, 412(3), 777-793 (2019-12-21)
Stratum corneum lipids are responsible for the skin's barrier function. They are the final product of epidermis lipid biosynthesis. During this process, lipids evolve from simple to complex structures in three main levels respectively (stratum basal level, stratum granulosum level
Nesli-Ece Sen et al.
International journal of molecular sciences, 20(23) (2019-11-27)
Ataxin-2 (human gene symbol ATXN2) acts during stress responses, modulating mRNA translation and nutrient metabolism. Ataxin-2 knockout mice exhibit progressive obesity, dyslipidemia, and insulin resistance. Conversely, the progressive ATXN2 gain of function due to the fact of polyglutamine (polyQ) expansions
Maria Tikhonenko et al.
PloS one, 8(1), e55177-e55177 (2013-02-06)
The vasodegenerative phase of diabetic retinopathy is characterized by not only retinal vascular degeneration but also inadequate vascular repair due to compromised bone marrow derived endothelial progenitor cells (EPCs). We propose that n-3 polyunsaturated fatty acid (PUFA) deficiency in diabetes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service