Skip to Content
Merck
All Photos(1)

Key Documents

860584P

Avanti

16:0 SM (d18:1/16:0)

Avanti Research - A Croda Brand, powder

Synonym(s):

N-palmitoyl-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H79N2O6P
CAS Number:
Molecular Weight:
703.03
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TOF, LC/MS)

form

powder

packaging

pkg of 1 × 10 mg (860584P-10mg)
pkg of 1 × 25 mg (860584P-25mg)
pkg of 1 × 5 mg (860584P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C39H79N2O6P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-38(42)37(36-47-48(44,45)46-35-34-41(3,4)5)40-39(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h30,32,37-38,42H,6-29,31,33-36H2,1-5H3,(H-,40,43,44,45)/b32-30+/t37-,38+/m0/s1

InChI key

RWKUXQNLWDTSLO-GWQJGLRPSA-N

Application

N-palmitoyl-D-erythro-sphingosylphosphorylcholine (16:0 SM (d18:1/16:0)) has been used as a standard for the quantitation of sphingolipid metabolites. It has also been used for the quantification of lipids in green fluorescence protein (GFP) and fatty acid transport protein 4 (FATP4) cells using electrospray ionization tandem mass spectrometry (ESI-MS/MS). It may be used in liposomes to measure ostreolysin A (OlyA) binding to determine the effects of phosphatidylcholines (PCs) on sphingomyelin (SM)/cholesterol interactions.

Packaging

5 mL Amber Glass Screw Cap Vial (860584P-10mg)
5 mL Amber Glass Screw Cap Vial (860584P-25mg)
5 mL Amber Glass Screw Cap Vial (860584P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabetta Albi et al.
International journal of molecular sciences, 15(7), 11555-11565 (2014-07-01)
Proliferating thyroid cells are more sensitive to UV-C radiations than quiescent cells. The effect is mediated by nuclear phosphatidylcholine and sphingomyelin metabolism. It was demonstrated that proton beams arrest cell growth and stimulate apoptosis but until now there have been
Shreya Endapally et al.
Cell, 176(5), 1040-1053 (2019-02-05)
Sphingomyelin and cholesterol are essential lipids that are enriched in plasma membranes of animal cells, where they interact to regulate membrane properties and many intracellular signaling processes. Despite intense study, the interaction between these lipids in membranes is not well
Ebru Afşar et al.
Redox biology, 30, 101430-101430 (2020-01-25)
We aimed to determine sphingolipid levels and examine apoptotic pathways in human retinal pigment epithelial cells (ARPE-19) undergoing endoplasmic reticulum (ER) stress. Cells were treated with tunicamycin (TM) to induce ER stress and tauroursodeoxycholic acid (TUDCA), an ER stress inhibitor
Xingzhong Zhang et al.
Cell metabolism, 30(5), 937-951 (2019-11-02)
Obesity-induced adipose dysfunction is a major contributor to atherosclerosis. Cold exposure has been reported to affect atherosclerosis through regulation of adipose function, but the mechanism has not been well clarified. Here, adipocyte hypoxia-inducible factor 2α (HIF-2α) was upregulated after mild cold
Jung Yong Eum et al.
Journal of chromatography. A, 460849-460849 (2020-01-14)
Aging refers to the intracellular accumulation of reactive oxygen species that damages proteins, DNA, and lipids. As alterations in lipid metabolism may trigger metabolic disorders and the onset of metabolic diseases, changes in lipid profiles can be closely related to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service