Skip to Content
Merck
All Photos(1)

Key Documents

860850P

Avanti

CER1 (d18:1/26:0/18:1)

Avanti Research - A Croda Brand 860850P, powder

Synonym(s):

N-[26-oleoyloxy hexacosanoyl]-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H119NO5
CAS Number:
Molecular Weight:
958.61
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860850P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860850P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(N[C@H]([C@H](O)/C=C/CCCCCCCCCCCCC)CO)CCCCCCCCCCCCCCCCCCCCCCCCCOC(CCCCCCC/C=C\CCCCCCCC)=O

General description

CER1 (d18:1/26:0/18:1), also referred as N-[26-oleoyloxy hexacosanoyl]-D-erythro-sphingosine, is an esterified ceramide. This lipid contains esterified ω-hydroxy (EO) fatty acid and 2 hydroxyl groups on its base moiety.

Packaging

5 mL Amber Glass Screw Cap Vial (860850P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Assi et al.
Analytical and bioanalytical chemistry, 412(3), 777-793 (2019-12-21)
Stratum corneum lipids are responsible for the skin's barrier function. They are the final product of epidermis lipid biosynthesis. During this process, lipids evolve from simple to complex structures in three main levels respectively (stratum basal level, stratum granulosum level

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service