Skip to Content
Merck
All Photos(2)

Key Documents

860051P

Avanti

Ceramide (Egg)

Ceramide (Egg, Chicken), powder

Synonym(s):

C16 Ceramide; N-hexadecanoyl-D-erythro-sphingosine; N-(hexadecanoyl)-sphing-4-enine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (860051P-200mg)
pkg of 1 × 25 mg (860051P-25mg)
pkg of 1 × 500 mg (860051P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860051P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

N([C@H]([C@H](O)\C=C\CCCCCCCCCCCCC)CO)C(=O)CCCCCCCCCCCCCCC

InChI

1S/C34H67NO3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(37)32(31-36)35-34(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h27,29,32-33,36-37H,3-26,28,30-31H2,1-2H3,(H,35,38)/b29-27+/t32-,33+/m0/s1

InChI key

YDNKGFDKKRUKPY-TURZORIXSA-N

Application

Ceramide has been used as lipid standard and as a growth medium for human peripheral blood mononuclear cells (PBMCs).

Biochem/physiol Actions

Ceramide plays a major role in the anabolic and catabolic pathways of sphingolipids. It acts as a extracellular stimuli for many cellular processes like differentiation, growth suppression, cell senescence and apoptosis.

Packaging

20 mL Clear Glass Screw Cap Vial (860051P-500mg)
5 mL Amber Glass Screw Cap Vial (860051P-200mg)
5 mL Amber Glass Screw Cap Vial (860051P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The lipids C2-and C16-ceramide form large stable channels Implications for apoptosis
Siskind L J and Colombini M
The Journal of biological chemistry, 275(49), 38640-38644 (2000)
Invariant natural killer T cells from children with versus without food allergy exhibit differential responsiveness to milk-derived sphingomyelin
Jyonouchi S, et al.
The Journal of Allergy and Clinical Immunology, 128(1), 102-109 (2011)
Sphingomyelin synthase 1 suppresses ceramide production and apoptosis post-photodamage
Separovic D, et al.
Biochemical and Biophysical Research Communications, 358(1), 196-202 (2007)
LipidFrag: Improving reliability of in silico fragmentation of lipids and application to the Caenorhabditis elegans lipidome
Witting M, et al.
PLoS ONE, 12(3), e0172311-e0172311 (2017)
Optimizing a ultrahigh pressure liquid chromatography-time of flight-mass spectrometry approach using a novel sub-2 mum core-shell particle for in depth lipidomic profiling of Caenorhabditis elegans
Witting M, et al.
Journal of Chromatography A, 1359, 91-99 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service