Skip to Content

22244

Ceramide from bovine spinal cord

≥98.0% (TLC)

Synonym(s):

Bovine Spinal Cord Ceramide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine spinal cord

Quality Segment

assay

≥98.0% (TLC)

form

solid

impurities

α-hydroxy fatty acids

solubility

chloroform/methanol (99:1): 20 mg/mL, clear, colorless

lipid type

sphingolipids

shipped in

wet ice

storage temp.

−20°C

Application

Ceramide is used to investigate effects on programmed cell death (apoptosis) and insulin signaling.

Biochem/physiol Actions

Ceramide is a minor component of cell membranes. Activates atypical protein kinase C (e.g., PKCzeta) .

Preparation Note

from cerebrosides by a modification of the procedure by H.E. Carter et al., J. Lipid Res. 2, 228 (1961)

Other Notes

Sales restrictions may apply


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library