Skip to Content
Merck
All Photos(2)

Key Documents

857223P

Avanti

C16 LPA

1-O-hexadecyl-2-hydroxy-sn-glycero-3-phosphate (ammonium salt), powder

Synonym(s):

1-hexadecyl-sn-glycero-3-phosphate (ammonium salt); vPA(16:0e/0:0); 110683

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H44NO6P
CAS Number:
Molecular Weight:
413.53
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPA; may contain up to 10% of the 2-LPA isomer, TLC)

form

powder

packaging

pkg of 1 × 1 mg (857223P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857223P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(O)=O)([H])COCCCCCCCCCCCCCCCC.[NH4+]

General description

Lysophosphatidic acid (LPA) is a lysophospholipid (LPL), that is made up of glycerol 3-phosphate (G3P), fatty acids, and phosphates. It is mainly produced in plants.

Biochem/physiol Actions

Lysophosphatidic acid (LPA) participates in the synthesis of phosphatidic acid (PA).

Packaging

5 mL Amber Glass Screw Cap Vial (857223P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bioactive lipids in gintonin-enriched fraction from ginseng
Cho HJ, et al.
Journal of ginseng research (2017)
Jesica R Williams et al.
The Journal of biological chemistry, 284(25), 17304-17319 (2009-04-16)
Lysophosphatidic acid (LPA) is a ligand for LPA(1-3) of the endothelial differentiation gene family G-protein-coupled receptors, and LPA(4-8) is related to the purinergic family G-protein-coupled receptor. Because the structure-activity relationship (SAR) of GPR92/LPA(5) is limited and whether LPA is its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service