Skip to Content
Merck
All Photos(2)

Key Documents

840475C

Avanti

18:1 (Δ9-Cis) PG

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); DOPG; PG(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H78O10PNa
CAS Number:
Molecular Weight:
797.03
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dioleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform solution

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840475C-25mg)
pkg of 2 × 4 mL (840475C-200mg)
pkg of 5 × 4 mL (840475C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840475C-25mg)
25 mg/mL (840475C-200mg)
25 mg/mL (840475C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C42H79O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,39-40,43-44H,3-16,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b19-17-,20-18-;/t39?,40-;/m

InChI key

IUVFCFQZFCOKRC-IPKKNMRRSA-M

General description

Phosphoglyceride (PG) are also referred as glycerophospholipid. It is an important component of membrane bilayers. It has three-carbon backbone of glycerol, two long-chain fatty acids, esterified to hydroxyl groups on carbons 1 and 2 of the glycerol and phosphoric acid esterified to the C3 hydroxyl group of glycerol. It is found at high levels in the membranes of all cells and at low levels in fat stores. 1,2-dioleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (DOPG)/18:1 (Δ9-Cis) PG has two hydroxyl groups in its head group. It is an anionic lipid.

Application


  • Probing the extended lipid anchorage with cytochrome c and liposomes containing diacylphosphatidylglycerol lipids.: This study investigates the interactions of cytochrome c with liposomes containing diacylphosphatidylglycerol (PG) lipids, including 18:1 (Δ9-Cis) PG. The research provides insights into the lipid anchorage and membrane dynamics, which are crucial for understanding cellular processes and developing biotechnological applications involving lipid-protein interactions (Abbott et al., 2018).

Biochem/physiol Actions

Phosphoglycerides (PG)/glycerophospholipids play a key role in cell signaling systems. It also serves as an anchor for proteins in cell membranes.

Packaging

5 mL Clear Glass Sealed Ampule (840475C-200mg)
5 mL Clear Glass Sealed Ampule (840475C-25mg)
5 mL Clear Glass Sealed Ampule (840475C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane Structure and Dynamics
Cell Biology, 227- 239 (2017)
Lipids
Medical Biochemistry, 99- 119 (2017)
Choosing the right fluorophore for single-molecule fluorescence studies in a lipid environment
Zhang Z, et al.
Biochimica et Biophysica Acta - Biomembranes, 1859(7), 1242- 1253 (2017)
Mariya Chavarha et al.
Biophysical journal, 104(3), 594-603 (2013-02-28)
The hydrophobic surfactant proteins, SP-B and SP-C, greatly accelerate the adsorption of the surfactant lipids to an air/water interface. Previous studies of factors that affect curvature suggest that vesicles may adsorb via a rate-limiting structure with prominent negative curvature, in
Md Zahidul Islam et al.
Langmuir : the ACS journal of surfaces and colloids, 33(9), 2433-2443 (2017-02-07)
The translocation of cell-penetrating peptides (CPPs) through plasma membranes of living cells is an important physiological phenomenon in biomembranes. To reveal the mechanism underlying the translocation of a CPP, transportan 10 (TP10), through lipid bilayers, we examined the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service