Skip to Content
Merck
All Photos(2)

Key Documents

810161P

Avanti

14:0-06:0 NBD PG

Avanti Research - A Croda Brand 810161P, powder

Synonym(s):

1-myristoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H56N5O13P
CAS Number:
Molecular Weight:
749.79
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810161P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810161P

shipped in

dry ice

storage temp.

−20°C

General description

7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) phosphatidylglycerol (PG) is a fluorescently labeled lipid where the NBD moiety is attached to the phosphatidylglycerol (PG). PG is abundant in the pulmonary surfactant. It is localized in the alveoli.

Application

16:0-06:0 NBD PG is suitable for lipoteichoic acid synthase (eLtaS) enzyme assay.

Biochem/physiol Actions

7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) fluorescent labeled lipids function as potential fluorophores for the study of biological and model membranes. 7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) phosphatidylglycerol (PG) is useful in the analysis of enzymatic reactions. Phosphatidylglycerol (PG) controls inflammatory mechanism against viral infections.

Packaging

5 mL Amber Glass Screw Cap Vial (810161P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi-Jung Weng et al.
Molecular membrane biology, 33(1-2), 23-28 (2016-07-28)
To investigate the effect of fluorescent probe on the properties of membranes, we studied model membranes composed of 1,2- dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) and 1-palmitoyl 2-oleoyl-sn-glycero-3-phosphocholine (POPC) in the presence and absence of fluorescent probe. The morphology of giant unilamellar vesicles (GUVs)
H Raghuraman et al.
Biochimica et biophysica acta, 1768(5), 1258-1267 (2007-03-17)
Lipids that are labeled with the NBD (7-nitrobenz-2-oxa-1,3-diazol-4-yl) group are widely used as fluorescent analogues of native lipids in biological and model membranes to monitor a variety of processes. The NBD group of acyl chain labeled NBD lipids is known
Maria Mercedes Palomino et al.
Microbiology (Reading, England), 159(Pt 11), 2416-2426 (2013-09-10)
The probiotic Gram-positive bacterium Lactobacillus casei BL23 is naturally confronted with salt-stress habitats. It has been previously reported that growth in high-salt medium, containing 0.8 M NaCl, leads to modifications in the cell envelope of this bacterium. In this study
Rachel Fickes et al.
Rapid communications in mass spectrometry : RCM, 30(24), 2601-2606 (2016-10-01)
Structural analogs of the bioactive lipid 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoglycerol were synthesized with a xylitol polar head group and both diacyl and diether radyl groups. Mass spectral characterization of xylitol phospholipids (PX) was carried out using collisional activation and high-resolution mass measurements of
Mirka E Wörmann et al.
Molecular microbiology, 79(3), 566-583 (2011-01-25)
Lipoteichoic acid (LTA) is an important cell wall polymer in Gram-positive bacteria. The enzyme responsible for polyglycerolphosphate LTA synthesis is LtaS, first described in Staphylococcus aureus. Four LtaS orthologues, LtaS(BS) , YfnI, YqgS and YvgJ, are present in Bacillus subtilis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service