Skip to Content
Merck
All Photos(2)

Key Documents

810153P

Avanti

16:0-06:0 NBD PE

Avanti Research - A Croda Brand 810153P, powder

Synonym(s):

1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H56N5O11P
CAS Number:
Molecular Weight:
729.80
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810153P-1mg)
pkg of 5 × 1 mg (810153P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810153P

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is a fluorescently labeled lipid localized at the membrane interface. The molecule contains NBD label covalently linked to the headgroup of phosphatidylethanolamine. PE is mostly present in the brain and is the second most abundant, metabolically related aminophospholipid.

Application

16:0-06:0 NBD PE is suitable for the determination of enzyme substrate specificity of lipoteichoic acid synthase (LtaS).

Biochem/physiol Actions

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is involved in screening the effects of Red edge excitation shift (REES) experiment with the unique motional and dielectric property it possesses. PE functions as a substrate for a number of phospholipids of the cell membrane.

Packaging

5 mL Amber Glass Screw Cap Vial (810153P-1mg)
5 mL Amber Glass Screw Cap Vial (810153P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of NBD-labeled lipids in membrane and cell biology
Fluorescent methods to study biological membranes, 37-50 (2012)
Jean E Vance et al.
Biochimica et biophysica acta, 1831(3), 543-554 (2012-09-11)
Phosphatidylserine (PS) and phosphatidylethanolamine (PE) are metabolically related membrane aminophospholipids. In mammalian cells, PS is required for targeting and function of several intracellular signaling proteins. Moreover, PS is asymmetrically distributed in the plasma membrane. Although PS is highly enriched in
Maria Karatsa-Dodgson et al.
Journal of bacteriology, 192(20), 5341-5349 (2010-08-17)
Lipoteichoic acid (LTA) is an important cell wall component of Gram-positive bacteria. The key enzyme responsible for polyglycerolphosphate lipoteichoic acid synthesis in the Gram-positive pathogen Staphylococcus aureus is the membrane-embedded lipoteichoic acid synthase enzyme, LtaS. It is presumed that LtaS
S C Lopes et al.
Biochimica et biophysica acta. Biomembranes, 1861(6), 1152-1161 (2019-03-07)
Mitochondrial membranes are pointed out as the site of cardiotoxic action of local anaesthetics. Its three main phospholipids components are phosphatidylcholine, phosphatidylethanolamine and cardiolipin. Cardiolipins, in eukaryotes, are only found in mitochondria and are essential for the maintenance of its
Maria Mercedes Palomino et al.
Microbiology (Reading, England), 159(Pt 11), 2416-2426 (2013-09-10)
The probiotic Gram-positive bacterium Lactobacillus casei BL23 is naturally confronted with salt-stress habitats. It has been previously reported that growth in high-salt medium, containing 0.8 M NaCl, leads to modifications in the cell envelope of this bacterium. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service