Skip to Content
Merck
All Photos(1)

Key Documents

710332C

Avanti

14:0 Cardiolipin (ammonium salt)

1′,3′-bis[1,2-dimyristoyl-sn-glycero-3-phospho]-glycerol (ammonium salt), chloroform

Synonym(s):

1,1′,2,2′-tetramyristoyl cardiolipin (ammonium salt); 1,1′,2,2′-tetratetradecanoyl cardiolipin (ammonium salt); CL(1′-[14:0/14:0],3′-[14:0/14:0])

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H132N2O17P2
CAS Number:
Molecular Weight:
1275.69
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

solution

packaging

pkg of 1 × 2.5 mL (710332C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 710332C

concentration

10 mg/mL (710332C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

14:0 Cardiolipin is an acidic phospholipid. Structurally, cardiolipin comprises three glycerol moieties and four acyl chains and is localized in the inner membrane of mitochondria. 14:0 Cardiolipin contains four myristic acid as acyl chain.

Application

14:0 Cardiolipin (1′,3′-bis[1,2-dimyristoyl-sn-glycero-3-phospho]-glycerol (ammonium salt)) has been used:
  • as a standard in tandem mass spectrometry
  • in the preparation of membrane proximal region (MPR) of human immunodeficiency virus (HIV-1) glycoprotein 41 (gp41)-lipopeptide conjugate and liposomes
  • as a component of micelle for suspending C8 ceramide substrate in ceramide kinase assay

Biochem/physiol Actions

Cardiolipin (CL) plays a key role in oxidative phosphorylation. CL is essential for energy production and in the activation of apoptosis. Deficiency of CL is associated with the development of diabetes and age-related heart failure. Anomalies associated with CL composition is observed in Barth Syndrome (BTHS).

Packaging

5 mL Clear Glass Sealed Ampule (710332C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ninus Simonzadeh
Journal of chromatographic science, 47(4), 304-308 (2009-05-02)
Phospholipids, such as 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC), and 1,1',2,2'-tetramyristoyl cardiolipin, along with cholesterol, form liposomes in aqueous media and have been investigated at NeoPharm (Lake Bluff, IL) as drug-delivery systems. To accurately assess the effectiveness of various formulations involving the use of
Anja Stulz et al.
Langmuir : the ACS journal of surfaces and colloids, 35(49), 16366-16376 (2019-11-12)
Most antimicrobial peptides (AMPs) and their synthetic mimics (SMAMPs) are thought to act by permeabilizing cell membranes. For antimicrobial therapy, selectivity for pathogens over mammalian cells is a key requirement. Understanding membrane selectivity is thus essential for designing AMPs and
Fang-Cheng Bi et al.
The Plant cell, 26(8), 3449-3467 (2014-08-26)
Arabidopsis thaliana plants that lack ceramide kinase, encoded by ACCELERATED CELL DEATH5 (ACD5), display spontaneous programmed cell death late in development and accumulate substrates of ACD5. Here, we compared ceramide accumulation kinetics, defense responses, ultrastructural features, and sites of reactive
Anastacia M Garcia et al.
American journal of physiology. Heart and circulatory physiology, 318(4), H787-H800 (2020-02-15)
Despite advances in both medical and surgical therapies, individuals with single ventricle heart disease (SV) remain at high risk for the development of heart failure (HF). However, the molecular mechanisms underlying remodeling and eventual HF in patients with SV are
Joanna Juhaniewicz-Dębińska et al.
Langmuir : the ACS journal of surfaces and colloids, 36(19), 5324-5335 (2020-04-29)
Daptomycin is known as an effective antibiotic lipopeptide which shows activity against the number of Gram-positive pathogens. Its primary target is the bacterial cell membrane. However, the detailed mechanism of daptomycin action is still subject to debate. In this paper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service