Skip to Content
Merck
All Photos(1)

Key Documents

710332P

Avanti

14:0 Cardiolipin (ammonium salt)

Avanti Research - A Croda Brand

Synonym(s):

1,1′,2,2′-tetramyristoyl cardiolipin (ammonium salt); 1,1′,2,2′-tetratetradecanoyl cardiolipin (ammonium salt); CL(1′-[14:0/14:0],3′-[14:0/14:0])

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H132N2O17P2
CAS Number:
Molecular Weight:
1275.69
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1′,3′-bis[1,2-dimyristoyl-sn-glycero-3-phospho]-glycerol (ammonium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (710332P-200mg)
pkg of 1 × 25 mg (710332P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

14:0 Cardiolipin or TMCL (1,1′,2,2′-tetramyristoyl cardiolipin) is a saturated CL and exists as gel at low temperatures.
Cardiolipin is an anionic phospholipid. The 14:0 indicates the fatty acyl substituents attached to the carbons of glycerol. It is localized to the inner mitochondrial membrane.

Application

14:0 Cardiolipin (ammonium salt) has been used as stable isotope-labeled standard prior to yeast strains high performance liquid chromatography electrospray ionization-tandem mass spectrometry (HPLC-ESI-MS/MS). It may be used as an internal standard in mass spectroscopy and in liposome preparation for electrophysiological studies.
14:0 Cardiolipin (ammonium salt) may be used in lipopeptide synthesis.

Biochem/physiol Actions

Cardiolipin participates in proton uptake pathway and also maintains the structure of enzymes involved. Cardiolipin maintains the mitochondrial function, membrane potential and structural support for the inner mitochondrial membrane and proteins in it. Concentration and composition changes of cardiolipin have been implicated in pathological conditions including ischemia, hypothyroidism, aging, heart failure and cardioskeletal myopathy.) Cardiolipin can form micelles/vesicles naturally.
TMCL (1,1′,2,2′-tetramyristoyl cardiolipin) bilayers are useful in generating multilamellar vesicles. Defects in the cardiolipin metabolic associated with Barth syndrome patients. Liposomes of TMCL with 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) and cholesterol are potential drug-delivery systems. TMCL displays temperature dependent thermotropic phase transitions.

Packaging

5 mL Amber Glass Screw Cap Vial (710332P-200mg)
5 mL Amber Glass Screw Cap Vial (710332P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC/MS analysis of cardiolipins in substantia nigra and plasma of rotenone-treated rats: implication for mitochondrial dysfunction in Parkinson's disease
Tyurina YY, et al.
Free Radical Research, 49(5), 681-691 (2015)
Role of lipid structure in the humoral immune response in mice to covalent lipid-peptides from the membrane proximal region of HIV-1 gp41
Watson DS and Szoka FC
Vaccine, 27(34), 4672-4683 (2009)
CLD1 Reverses the Ubiquinone Insufficiency of Mutant cat5/coq7 in a Saccharomyces cerevisiae Model System
Kar A, et al.
Testing, 11(9), e0162165-e0162165 (2016)
Assessing the effect of the lipid environment on the redox potentials of the coenzymes Q10 and Q4 using lipid monolayers made of DOPC, DMPC, TMCL, TOCL, and natural cardiolipin (nCL) on mercury
Heise N and Scholz F
Electrochemical Communications, 81, 141-144 (2017)
Interactions of Monovalent and Divalent Cations with Cardiolipin Monolayers
Kensbock R, et al.
Langmuir, 35(10), 3624-3633 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service