Skip to Content
Merck
All Photos(2)

Key Documents

700025P

Avanti

7β,27-dihydroxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

(3β,7β,25R)-Cholest-5-ene-3,7,26-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O3
CAS Number:
Molecular Weight:
418.65
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (700025P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=C[C@@H]2O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC[C@@H](C)CO

InChI

1S/C27H46O3/c1-17(16-28)6-5-7-18(2)21-8-9-22-25-23(11-13-27(21,22)4)26(3)12-10-20(29)14-19(26)15-24(25)30/h15,17-18,20-25,28-30H,5-14,16H2,1-4H3/t17-,18-,20+,21-,22+,23+,24+,25+,26+,27-/m1/s1

InChI key

RXMHNAKZMGJANZ-BMOLSTJGSA-N

General description

7β,27-dihydroxycholesterol is a substrate for enzyme 11β-hydroxysteroid dehydrogenases (11β-HSDs).

Application

7β,27-dihydroxycholesterol has been used as an internal standard in ultra-performance liquid chromatography -tandem mass spectrometer (UPLC-MS/MS). It has also been used as a substrate for 11β-hydroxysteroid dehydrogenases (11β-HSDs) in human embryonic kidney cells (HEK293).

Biochem/physiol Actions

7β,27-dihydroxycholesterol (7β27OHC) is a retinoid-related orphan receptor γ (RORγ) ligand. 7β27OHC favors smoothened (Smo) functionality.

Packaging

5 mL Amber Glass Screw Cap Vial (700025P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

11beta-Hydroxysteroid dehydrogenases control access of 7beta, 27-dihydroxycholesterol to retinoid-related orphan receptor gamma
Beck KR, et al.
Journal of Lipid Research, 60(9), 1535-1546 (2019)
Assessment of Altered Cholesterol Homeostasis by Xenobiotics Using Ultra-High Performance Liquid Chromatography-Tandem Mass Spectrometry
Herron J, et al.
Current Protocols in Toxicology / Editorial Board, Mahin D. Maines (editor-in-chief) ... [Et al.], 78(1), e65-e65 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service