Z743731
ART® Low Retention Barrier Tip
ART 1000 REACH, size 1000 μL, hinged rack, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
About This Item
hinged rack
Recommended Products
sterility
sterile
feature
ART 1000 REACH
hinged rack
packaging
pkg of 4X768 ea
manufacturer/tradename
Molecular BioProducts 2079-05-HR
technique(s)
PCR: suitable
size
1000 μL
volume
1000 μL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- ART low retention barrier tips ensure complete protection against aerosols and liquid contamination.
- The tips are free from DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
- Stackable barrier tip packaging enables tips to pass through offset holes in the lower insert.
- Blister sleeve guide rails facilitate smooth transfer of tip inserts into the rack.
- An outer sleeve shields tips from external contamination during the refilling of inserts.
- The tip packaging option helps reduce plastic waste.
- The rigid insert design ensures reliable tip loading.
- Lot number and description are printed on every insert for easy tracking and identification.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service