Skip to Content
Merck
All Photos(1)

Key Documents

CLS4140

Corning® DeckWorks barrier pipet tips

100 - 1000μL low bind barrier pipet tips, extended length, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 8 racks/pack, 4 packs/case

Synonym(s):

Pipette tips, low binding barrier pipet tips, low binding barrier pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02

material

natural tip
polypropylene

sterility

sterile

feature

DNase free
RNase free
barrier
disposable
endotoxin free
filter
graduated
Tip shape: round

packaging

case of 3840
rack of 96

manufacturer/tradename

Corning 4140

volume range

100-1000 μL

suitability

suitable for (quantitative and semi-quantitative assays such a real-time PCR (qPCR) and conventional PCR)

binding type

low binding surface

General description

Corning® DeckWorks Barrier Tips

Corning DeckWorks barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.

This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.

The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.

Medical grade resins and production excellence allow for full certification as:

  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free
  • Sterility Assurance Level 10-3

Legal Information

Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service