Skip to Content
Merck
All Photos(1)

Key Documents

W501808

Sigma-Aldrich

5-(Hydroxymethyl)furfural

≥99%, FG

Synonym(s):

5-Hydroxymethyl-2-furaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethyl-2-furancarboxaldehyde, HMF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
110889
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.139
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

Assay

≥99%

refractive index

n20/D 1.562 (lit.)

bp

114-116 °C/1 mmHg (lit.)

mp

28-34 °C (lit.)

density

1.243 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

butter; caramel; fatty; musty; waxy

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(CO)o1

InChI

1S/C6H6O3/c7-3-5-1-2-6(4-8)9-5/h1-3,8H,4H2

InChI key

NOEGNKMFWQHSLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Resultant compound from sublimation test for Gentianae Radix in Japanese Pharmacopoeia was 5-(hydroxymethyl)furfural.: This letter to the editor confirms the presence of 5-(Hydroxymethyl)furfural in traditional medicine preparations, discussing its implications for quality control and safety standards in pharmaceutical contexts (Tsuge et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yin-Ying Lee et al.
Physical chemistry chemical physics : PCCP, 14(40), 13914-13917 (2012-09-20)
Mesoporous silica nanoparticles functionalized with both sulfonic acid (HSO(3)) and ionic liquid (ILs) were synthesized and applied as effective and recyclable catalysts for generating 5-hydroxymethylfurfural (HMF) from fructose. For the first time a high HMF yield of 72.5% was achieved
Arvind H Jadhav et al.
Bioresource technology, 132, 342-350 (2013-02-26)
Acidity modified silver exchanged silicotungstic acid (AgSTA) catalyst was prepared and characterized by X-ray diffraction, FT-IR spectroscopy, Raman spectroscopy, FT-IR pyridine adsorption, SEM imaging, EDX mapping, and antimicrobial activity was also tested. The catalytic activity was evaluated for the dehydration
Monika Krystof et al.
ChemSusChem, 6(4), 630-634 (2013-03-05)
5-Hydroxymethylfurfural (HMF) is a valuable biomass-derived building block. Among possible HMF valorization products, a broad range of HMF esters can be synthesized. These HMF esters have found some promising applications, such as monomers, fuels, additives, surfactants, and fungicides, and thus
Lilong Zhou et al.
Bioresource technology, 129, 450-455 (2012-12-26)
A new kind of bifunctional ionic liquid catalysts was synthesized to degrade microcrystalline cellulose in [BMIM]Cl at atmospheric pressure. The effects of reaction temperature, amount of catalysts, reaction time, ionic liquid purity and cellulose concentration on conversion were investigated. At
Tim Ståhlberg et al.
ChemSusChem, 4(4), 451-458 (2011-01-29)
The synthesis of 5-(hydroxymethyl)furfural (HMF) in ionic liquids is a field that has grown rapidly in recent years. Unique dissolving properties for crude biomass in combination with a high selectivity for HMF formation from hexose sugars make ionic liquids attractive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service