Skip to Content
Merck
All Photos(1)

Documents

W270202

Sigma-Aldrich

5-Methylfurfural

≥98%, FG

Synonym(s):

5-Methyl-2-furaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
FEMA Number:
2702
Beilstein:
106895
EC Number:
Council of Europe no.:
119
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.001
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

impurities

<0.1% DMF (residual starting material)

refractive index

n20/D 1.531 (lit.)

bp

187 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; maple; spicy; sweet

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(C)o1

InChI

1S/C6H6O2/c1-5-2-3-6(4-7)8-5/h2-4H,1H3

InChI key

OUDFNZMQXZILJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methylfurfural has been identified as one of the volatile flavor compounds in tamarind, Dutch Coffee, barrel-aged wines and deep fat-fried soybeans.

Application


  • Metabolomics fingerprint of Philippine coffee by SPME-GC-MS for geographical and varietal classification.: This study employs solid-phase microextraction gas chromatography-mass spectrometry (SPME-GC-MS) to create a metabolomics fingerprint of Philippine coffee, identifying compounds like 5-Methylfurfural for the classification of coffee by geography and variety. This research highlights the potential of 5-Methylfurfural in distinguishing coffee varieties, showcasing its application in food biochemistry (Ongo et al., 2020).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile flavor components of deep fat-fried soybeans.
Wilkens WF & Lin FM.
Journal of Agricultural and Food Chemistry, 18, 337-339 (1970)
Volatile flavor components of tamarind (Tamarindus indica L.).
Zhang Y, et al.
J. Essent. Oil Res., 2(4), 197-198 (1990)
Formation and degradation of furfuryl alcohol, 5-methylfurfuryl alcohol, vanillyl alcohol, and their ethyl ethers in barrel-aged wines.
Spillman PJ, et al.
Journal of Agricultural and Food Chemistry, 46(2), 657-663 (1998)
Studies on the Flavor Compounds of Dutch Coffee by Headspace GC-Mass.
Hwang SH, et al.
Korean journal of food and cookery science, 30(5), 596-602 (2014)
Nick Wierckx et al.
Microbial biotechnology, 3(3), 336-343 (2011-01-25)
The formation of toxic fermentation inhibitors such as furfural and 5-hydroxy-2-methylfurfural (HMF) during acid (pre-)treatment of lignocellulose, calls for the efficient removal of these compounds. Lignocellulosic hydrolysates can be efficiently detoxified biologically with microorganisms that specifically metabolize the fermentation inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service