Skip to Content
Merck
All Photos(3)

Key Documents

W373011

Sigma-Aldrich

L-Rhamnose

greener alternative

natural sourced, 99%, FG

Synonym(s):

L-Rhamnose, 6-Deoxy-L-mannose, 6-Deoxyhexopyranose, 6-Deoxymannose

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H12O5 .H2O
CAS Number:
Molecular Weight:
182.17
FEMA Number:
3730
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

grade

FG
Halal
Kosher

Quality Level

reg. compliance

EU Regulation 178/2002

Assay

99%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

mp

90-95 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

odorless

SMILES string

O=C[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C.[H]O[H]

InChI

1S/C6H12O5.H2O/c1-3(8)5(10)6(11)4(9)2-7;/h2-6,8-11H,1H3;1H2/t3-,4-,5-,6-;/m0./s1

InChI key

CBDCDOTZPYZPRO-DEZHIRTDSA-N

General description

L-Rhamnose is a deoxy sugar mainly found in gums and plant mucilages. It is one of the structural units of complex polysaccharides such as pectin and sterculia gum. L-Rhamnose may react with aqueous ammonia to form volatile flavor compounds such as alkyl pyrazines.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudius Marondedze et al.
Frontiers in molecular biosciences, 6, 163-163 (2020-02-11)
The spliceosome processes RNAs from a pre-RNA state to a mature mRNA thereby influencing RNA availability for translation, localization, and turnover. It consists of complex structures containing RNA-binding proteins (RBPs) essential for post-transcriptional gene expression control. Here we investigate the
Jun Liu et al.
Carbohydrate polymers, 172, 11-19 (2017-06-14)
Nanocellulose-based biomaterials for biomedical and pharmaceutical applications have been extensively explored. However, studies on different levels of impurities in the nanocellulose and their potential risks are lacking. This article is the most comprehensive to date survey of the importance and
Formation of heterocyclic compounds from the reaction of L-rhamnose with ammonia.
Shibamoto T & Bernhard RA
Journal of Agricultural and Food Chemistry, 26(1), 183-187 (1978)
Adila Aipire et al.
Biomolecules, 10(1) (2020-01-23)
Glycyrrhiza uralensis is a Chinese herbal medicine with various bioactivities. Three fractions (GUPS-I, GUPS-II and GUPS-III) of G. uralensis polysaccharides (GUPS) were obtained with molecular weights of 1.06, 29.1, and 14.9 kDa, respectively. The monosaccharide compositions of GUPS-II and GUPS-III
Encyclopedia of Food and Color Additives, 1, 2583-2583 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service