Skip to Content
Merck
All Photos(2)

Key Documents

W312401

Sigma-Aldrich

Vanillylacetone

≥96%, FG

Synonym(s):

Zingerone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CH2CH2COCH3
CAS Number:
Molecular Weight:
194.23
FEMA Number:
3124
EC Number:
Council of Europe no.:
139
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.005
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 184.1099

Assay

≥96%

refractive index

n20/D 1.541 (lit.)

bp

141 °C/0.5 mmHg (lit.)

mp

40-41 °C (lit.)

density

1.14 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

ginger; woody; spicy; phenolic; sweet; vanilla

storage temp.

2-8°C

SMILES string

COc1cc(CCC(C)=O)ccc1O

InChI

1S/C11H14O3/c1-8(12)3-4-9-5-6-10(13)11(7-9)14-2/h5-7,13H,3-4H2,1-2H3

InChI key

OJYLAHXKWMRDGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Vanillylacetone is one of the key aroma constituents of ginger. It is a pharmacologically active compound that may contribute to the anti-inflammatory, anti-oxidant, anti-cancerous, radioprotective and anti-microbial properties of ginger.

Application

Vanillylacetone can be used as a food additive in food industries because of its sweet and spicy flavor.

Biochem/physiol Actions

Taste at 20 ppm in 5%

Other Notes

Natural occurrence: Cranberry and ginger.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mi Kyung Kim et al.
Experimental gerontology, 45(6), 419-426 (2010-03-10)
Zingerone, a major component found in ginger root, has been known as anti-mutagenic and anti-carcinogenic activities that are often associated with its anti-oxidative and anti-inflammatory activities. In recent studies, we examined molecular mechanism of zingerone treatment on pro-inflammatory NF-kappaB activation
Sang Woon Chung et al.
Journal of medicinal food, 12(2), 345-350 (2009-05-23)
Peroxisome proliferator-activated receptors (PPARs), members of the nuclear hormone receptor family, are key regulators of various metabolic pathways related to lipid and glucose metabolism as well as inflammation. We examined the effect of zingerone, a major ingredient of ginger, on
Mark A Affeltranger et al.
Chemical senses, 32(5), 455-462 (2007-04-03)
Sequential presentation of 2 irritants may produce cross-sensitization or cross-adaptation effects upon introduction of the second irritant. In Experiment 1, subjects were given either 34 min of stimulation with zingerone, capsaicin, or piperine or one of those irritants for 23
Fatih Mehmet Kandemir et al.
Environmental science and pollution research international, 26(22), 22562-22574 (2019-06-06)
Zingerone (ZO), one of the active components of ginger (Zingiber officinale), is a phenolic alkanone with antioxidant, antiapoptotic, and anti-inflammatory properties. Cisplatin (CP) is a widely used chemotherapeutic drug for solid tumors, but its therapeutic use is limited due to
Recent update on multiple pharmacological benefits of zingerone: a quick review.
Kumar L, et al.
American Journal of Phytomedicine and Clinical Therapeutics, 2, 693-704 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service