Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vanillylacetone
Synonym(s):
Zingerone
Linear Formula:
4-(HO)C
6
H
3
-3-(OCH
3
)CH
2
CH
2
COCH
3
CAS Number:
122-48-5
Molecular Weight:
194.23
EC Number:
204-548-3
Products
Product Category
aroma chemicals (2)
Brand
Sigma-Aldrich (2)
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Food Grade (2)
Fragrance grade (2)
Halal (2)
Kosher (1)
Natural (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
W312401
Vanillylacetone,
≥96%, FG
Expand
View Pricing
W312420
Vanillylacetone,
≥98%, natural (US), FG
Expand
View Pricing