Skip to Content
Merck
All Photos(1)

Key Documents

W309303

Sigma-Aldrich

2-Undecanone

≥98%, FCC, FG

Synonym(s):

Methyl nonyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COCH3
CAS Number:
Molecular Weight:
170.29
FEMA Number:
3093
Beilstein:
1749573
EC Number:
Council of Europe no.:
150
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.016
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

5.9 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.43 (lit.)

bp

231-232 °C (lit.)

mp

11-13 °C (lit.)

density

0.825 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; waxy; fruity

SMILES string

CCCCCCCCCC(C)=O

InChI

1S/C11H22O/c1-3-4-5-6-7-8-9-10-11(2)12/h3-10H2,1-2H3

InChI key

KYWIYKKSMDLRDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Undecanone occurs naturally in wild tomato. It is also found in Ruta chalepensis essential oil and hop oil. 2-Undecanone is an insect and tick repellant.

Application

  • (14)C-Isotope Use to Quantify Covalent Reactions between Flavor Compounds and β-Lactoglobulin.: Research utilizing 2-Undecanone to study its binding interactions with milk proteins, enhancing understanding of flavor compound stability and protein binding in food products (Shepelev and Reineccius, 2024).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2?Undecanone, a constituent of the glandular trichomes of Lycopersicon hirsutum f. glabratum: Effects on Heliothis zea and Manduca sexta growth and survival.
Farrar RR & Kennedy GG.
Entomologia Experimentalis et Applicata, 43(1), 17-23 (1987)
Constituents of the essential oil of Ruta chalepensis L. from Turkey.
Baser KHC, et al.
J. Essent. Oil Res., 8(4), 413-414 (1996)
Release characteristics of the non-toxic insect repellant 2-undecanone from its crystalline inclusion compound with ?-cyclodextrin.
Whang HS & Tonelli A.
Journal of Inclusion Phenomena and Macrocyclic Chemistry, 62(1-2), 127-134 (2008)
Novel arthropod repellent, BioUD, is an efficacious alternative to deet.
Witting-Bissinger BE, et al.
Journal of Medical Entomology, 45(5), 891-898 (2008)
Jing Chen et al.
International journal of molecular sciences, 15(12), 22978-22994 (2014-12-17)
Houttuynia cordata Thunb. (H. cordata) is an anti-inflammatory herbal drug that is clinically used in Asia. The essential oil obtained from H. cordata is known to contain 2-undecanone (2-methyl nonyl ketone). In addition, sodium houttuyfonate is a compound that can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service