Skip to Content
Merck
All Photos(1)

Documents

W254401

Sigma-Aldrich

2-Heptanone

≥98%, FCC, FG

Synonym(s):

Methyl pentyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COCH3
CAS Number:
Molecular Weight:
114.19
FEMA Number:
2544
Beilstein:
1699063
EC Number:
Council of Europe no.:
136
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.002
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

3.94 (vs air)

vapor pressure

2.14 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

739 °F

expl. lim.

1.11 %, 65 °F
7.9 %, 121 °F

refractive index

n20/D 1.408 (lit.)

bp

149-150 °C (lit.)

mp

−35 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

banana; creamy; fruity; waxy; cheesy

SMILES string

CCCCCC(C)=O

InChI

1S/C7H14O/c1-3-4-5-6-7(2)8/h3-6H2,1-2H3

InChI key

CATSNJVOTSVZJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Heptanone is a linear ketone. It has been produced in the cultures of Penicillium commune and Paecilomycet variotii cultivated on pine wood.
2-Heptanone is an aroma compound. It is a bioactive agent and has potential as a pesticide. Kinetics of release of 2-heptanone incorporated into lipid films or composite solid lipid particle (SLP) films have been investigated. 2-Heptanone is secreted by the mandibular glands of honeybees when they bite. It is reported to act as an anaesthetic in small arthropods, such as wax moth larva (WML) and Varroa mites, which are paralysed after a honeybee bite.

Application

2-Heptanone may be employed as solvent to determine the protection effectiveness of glove materials against solvents present in a commonly used automotive clear coat formulation using a novel permeation panel. It may be used in a study to investigate the influence of oxidation-induced structural modifications of myofibrillar protein on its binding ability with aroma compounds.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Bilbao-Sáinz et al.
Pest management science, 69(8), 975-982 (2013-01-29)
Controlled-release formulations of bioactive agents are of increasing interest for effective pest control. Volatile 2-heptanone is a bioactive agent that has shown potential as a pesticide. The aim of this study was to investigate the kinetics of release of 2-heptanone
Anna-Sara Claeson et al.
Journal of environmental monitoring : JEM, 4(5), 667-672 (2002-10-29)
Growth of different microorganisms is often related to dampness in buildings. Both fungi and bacteria produce complicated mixtures of volatile organic compounds that include hydrocarbons, alcohols, ketones, sulfur- and nitrogen-containing compounds etc. Microbially produced substances are one possible explanation of
Feibai Zhou et al.
Journal of agricultural and food chemistry, 62(39), 9544-9552 (2014-09-02)
The objective of this study was to investigate the influence of oxidation-induced structural modifications of myofibrillar protein on its binding ability with aroma compounds such as 2-methyl-butanal, methional, 2-pentanone, 2-heptanone, and nonanal. A method using solid-phase microextraction (SPME) combined with
Diana M Ceballos et al.
The Annals of occupational hygiene, 59(3), 358-372 (2014-12-03)
Determine protection effectiveness of 5-mil natural rubber latex (0.13-mm), 5-mil nitrile rubber (0.13-mm), and 13-mil butyl rubber (0.33-mm) glove materials against solvents present in a commonly used automotive clear coat formulation using a novel permeation panel. The latex and nitrile
Koutaroh Okada et al.
Chemical senses, 34(5), 425-433 (2009-04-14)
The olfactory system of an insect brain codes for information about odorant quality and quantity using the temporal pattern of neural activity as well as neurons' firing. Although an accurate odor temporal pattern is indispensable for investigations of olfactory systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service