Skip to Content
Merck
All Photos(1)

Key Documents

W303801

Sigma-Aldrich

Sucrose octaacetate

≥97%, FG

Synonym(s):

D-(+)-Sucrose octaacetate, Sucrose octaacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38O19
CAS Number:
Molecular Weight:
678.59
FEMA Number:
3038
Beilstein:
79290
EC Number:
Council of Europe no.:
4219
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.081
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515
FDA 21 CFR 175.105

Assay

≥97%

optical activity

[α]20/D +59°, c = 2.5 in ethanol

bp

260 °C (lit.)

mp

82-85 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

CC(=O)OC[C@H]1O[C@H](O[C@]2(COC(C)=O)O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@@H]2OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C28H38O19/c1-12(29)37-9-20-22(40-15(4)32)24(42-17(6)34)25(43-18(7)35)27(45-20)47-28(11-39-14(3)31)26(44-19(8)36)23(41-16(5)33)21(46-28)10-38-13(2)30/h20-27H,9-11H2,1-8H3/t20-,21-,22-,23-,24+,25-,26+,27-,28+/m1/s1

InChI key

ZIJKGAXBCRWEOL-SAXBRCJISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sucrose octaacetate is an acetylated analog of sucrose and a food-safe classical bitterant commonly used in food and dairy products.

Application


  • High-Degree Concentration Organic Solvent Forward Osmosis for Pharmaceutical Pre-Concentration.: Investigates the potential of organic solvent forward osmosis technologies in pharmaceutical applications, focusing on pre-concentration processes that may benefit from the stabilizing effects of sucrose octaacetate (Takada et al., 2024).

  • Nutrient-conditioned intake stimulation does not require a distinctive flavor cue in rats.: Explores behavioral responses in rats to various flavors including sucrose octaacetate, suggesting implications for understanding human dietary choices and the development of food additives (Sclafani et al., 2020).

  • Discrimination of Isointense Bitter Stimuli in a Beer Model System.: Examines the ability of individuals to distinguish between different bitter compounds, including sucrose octaacetate, in a controlled beer tasting model, which can influence food science and product development (Higgins and Hayes, 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Explaining tolerance for bitterness in chocolate ice cream using solid chocolate preferences
Harwood ML, et al.
Journal of Dairy Science, 96(8), 4938-4944 (2013)
Odor-cued bitter taste avoidance
Coppola DM and Slotnick B
Chemical Senses, 43(4), 239-247 (2018)
Meriel L Harwood et al.
Journal of food science, 77(10), S390-S393 (2012-08-29)
Classical detection thresholds do not predict liking, as they focus on the presence or absence of a sensation. Recently however, Prescott and colleagues described a new method, the rejection threshold, where a series of forced choice preference tasks are used
Jaehoon Kim et al.
Langmuir : the ACS journal of surfaces and colloids, 22(2), 642-657 (2006-01-13)
Ultrathin organic films of sucrose octaacetate (SOA) were deposited on 12.5 cm diameter silicon wafer substrates using high-pressure free meniscus coating (hFMC) with liquid CO2 (l-CO2) as a coating solvent. The dry film thickness across the wafer and the morphology
John D Boughter et al.
Chemical senses, 27(2), 133-142 (2002-02-13)
Inbred mouse strains vary in their response to bitter-tasting compounds as assessed by 48 h preference tests. These differences are generally assumed to result from altered gustatory function, although such long-term tests could easily reflect additional factors. We developed a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service