Skip to Content
Merck
All Photos(1)

Key Documents

W285803

Sigma-Aldrich

Phenethyl alcohol

≥99%, FCC, FG

Synonym(s):

2-Phenylethanol, β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2OH
CAS Number:
Molecular Weight:
122.16
FEMA Number:
2858
Beilstein:
1905732
EC Number:
Council of Europe no.:
68
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.019
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

4.21 (vs air)

vapor pressure

1 mmHg ( 58 °C)

Assay

≥99%

refractive index

n20/D 1.5317 (lit.)

bp

219-221 °C/750 mmHg (lit.)

mp

−27 °C (lit.)

density

1.020 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

honey; floral; rose

SMILES string

OCCc1ccccc1

InChI

1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

WRMNZCZEMHIOCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

recommended

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahra Seraj et al.
Scientific reports, 9(1), 18615-18615 (2019-12-11)
Numerous efforts have been directed towards investigating the different stages leading to the fibrillation process in neurodegenerative diseases and finding the factors modulating it. In this study, using a wide range of molecular techniques as well as fibrillation kinetics coupled
Jaspreet Singh et al.
Biochimica et biophysica acta, 1831(4), 747-758 (2013-01-16)
X-linked adrenoleukodystrophy (X-ALD) is a peroxisomal disorder caused by mutations in the ABCD1 gene. Accumulation of very long chain fatty acids (VLCFA) that have been attributed to reduced peroxisomal VLCFA β-oxidation activity are the hallmark of the disease. Overexpression of
Dongliang Hua et al.
Biotechnology advances, 29(6), 654-660 (2011-05-24)
2-Phenylethanol (2-PE) is an important aromatic alcohol with a rose-like fragrance. It has been widely applied in the cosmetic, perfume, and food industries and is mainly produced by chemical synthesis. An alternative method for the production of natural flavors and
M M W Etschmann et al.
Applied microbiology and biotechnology, 59(1), 1-8 (2002-06-20)
2-Phenylethanol (2-PE) is an important flavour and fragrance compound with a rose-like odour. Most of the world's annual production of several thousand tons is synthesised by chemical means but, due to increasing demand for natural flavours, alternative production methods are
G Pereira-Caro et al.
Food chemistry, 138(2-3), 1172-1182 (2013-02-16)
The anticarcinogenic activity of hydroxytyrosyl ethyl ether (HTy-Et) compared to its precursor hydroxytyrosol (HTy) has been studied in human Caco-2 colon adenocarcinoma cells. 451 and 977 genes were differentially expressed in Caco-2 cells exposed to HTy or HTy-Et for 24h

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service