Skip to Content
Merck
All Photos(1)

Key Documents

W243906

Sigma-Aldrich

Ethyl hexanoate

≥98%, FCC, FG

Synonym(s):

Caproic acid ethyl ester, Ethyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COOC2H5
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2439
Beilstein:
1701293
EC Number:
Council of Europe no.:
310
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.060
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

5 (vs air)

Assay

≥98%

refractive index

n20/D 1.407 (lit.)

bp

168 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana; green; waxy; fruity; pineapple; sweet

SMILES string

CCCCCC(=O)OCC

InChI

1S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3

InChI key

SHZIWNPUGXLXDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl hexanoate is the main flavor component identified in Chinese strong-flavored liquor (CSFL).

Application


  • Formation of Key Aroma Compounds During 30 Weeks of Ripening in Gouda-Type Cheese Produced from Pasteurized and Raw Milk.: The study investigates the formation of key aroma compounds, including ethyl hexanoate, during the ripening of Gouda-type cheese. The results provide insights into the ripening process and its impact on cheese flavor development (Duensing et al., 2024).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prokaryotic communities in pit mud from different-aged cellars used for the production of Chinese strong-flavored liquor.
Tao Yong, et al.
Applied and Environmental Microbiology, 80(7), 2254-2260 (2014)
Da-Mi Jung et al.
Journal of agricultural and food chemistry, 51(1), 200-205 (2002-12-28)
Changes in the volatility of selected flavor compounds in the presence of nonvolatile food matrix components were studied using headspace solid-phase microextraction (HS-SPME) combined with GC-MS quantification. Time-dependent adsorption profiles to the SPME fiber and the partition coefficients between different
Clémentine Lauverjat et al.
Journal of agricultural and food chemistry, 57(21), 9878-9887 (2009-10-13)
Physicochemical properties (partition and diffusion coefficients) involved in the mobility and release of salt and aroma compounds in model cheeses were determined in this study. The values of NaCl water/product partition coefficients highlighted interactions between proteins and NaCl. However, these
John C Beaulieu et al.
Journal of agricultural and food chemistry, 55(23), 9503-9513 (2007-10-20)
Changes in post-cutting volatiles, quality, and sensory attributes during fresh-cut storage (4 degrees C) of cantaloupe (Cucumis melo L. var. Reticulatus, Naudin, cv. 'Sol Real') harvested at four distinct maturities (1/4-, 1/2-, 3/4-, and full-slip) were investigated after 0, 2
Géraldine Savary et al.
Journal of agricultural and food chemistry, 55(17), 7099-7106 (2007-07-31)
The release of a strawberry aroma from different composite gels taken as models of fruit preparations and from a sucrose solution was investigated. The composition of the model systems differed with regard to the gelling agent, either pectin or carrageenan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service