Skip to Content
Merck
All Photos(1)

Key Documents

W279609

Sigma-Aldrich

γ-Octalactone

≥97%, FCC, FG

Synonym(s):

γ-Octanoic lactone, 4-Hydroxy octanoic acid, Octano-1,4-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
FEMA Number:
2796
EC Number:
Council of Europe no.:
2274
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.022
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.444 (lit.)

bp

234 °C (lit.)

density

0.981 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; creamy; peach; sweet

SMILES string

CCCCC1CCC(=O)O1

InChI

1S/C8H14O2/c1-2-3-4-7-5-6-8(9)10-7/h7H,2-6H2,1H3

InChI key

IPBFYZQJXZJBFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

γ-Octalactone is one of the flavor constituents of peaches, oranges and sweet fortified wines.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile components of peach.
Jennings WG & Sevenants MR.
Journal of Food Science, 29(6), 796-801 (1964)
I Cutzach et al.
Journal of agricultural and food chemistry, 47(7), 2837-2846 (1999-12-20)
Sweet fortified wines, traditionally aged under strong oxidation conditions, have a characteristic aroma. An experimental laboratory study investigated the aging of red and white sweet fortified wines under various conditions. The formation of various molecules, previously identified as characteristic of
Identification of aroma active compounds in orange essence oil using gas chromatography?olfactometry and gas chromatography?mass spectrometry.
Hognadottir A & Rouseff RL.
Journal of Chromatography A, 998(1), 201-211 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service