Skip to Content
Merck
All Photos(2)

Key Documents

W236004

Sigma-Aldrich

γ-Decalactone

≥98%, FCC, FG

Synonym(s):

γ-Decanolactone, 4-Hydroxydecanoic acid γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
FEMA Number:
2360
EC Number:
Council of Europe no.:
2230
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.017
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.449 (lit.)

bp

281 °C

density

0.948 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

creamy; fruity; peach; sweet

SMILES string

CCCCCCC1CCC(=O)O1

InChI

1S/C10H18O2/c1-2-3-4-5-6-9-7-8-10(11)12-9/h9H,2-8H2,1H3

InChI key

IFYYFLINQYPWGJ-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

γ-Decalactone, the main volatile aroma compound found in peaches, can be used as a flavoring agent in the food industry due to its distinct peach odor. It is also found in strawberries and apricots.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroma characterization of various apricot varieties using headspace?solid phase microextraction combined with gas chromatography?mass spectrometry and gas chromatography?olfactometry.
Guillot S, et al.
Food Chemistry, 96(1), 147-155 (2006)
Mohatmed Abdel-Latief et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(26), 8914-8919 (2008-06-27)
Epoxide hydrolases (EHs) are enzymes occurring in virtually any living organism. They catalyze the hydrolysis of epoxide containing lipids and are involved in crucial mechanisms, such as the detoxification of xenobiotics or the regulation of inflammation and blood pressure. Here
Scale-up of natural product formation and isolation.
Ralf G Berger et al.
Molecular nutrition & food research, 49(8), 732-743 (2005-07-02)
Hirotoshi Tamura et al.
Journal of agricultural and food chemistry, 53(13), 5397-5401 (2005-06-23)
Authenticity assessment of gamma-decalactone (1) and delta-decalactone (2) from peach (Prunus persica var. persica), apricot (Prunus armeniaca), and nectarine (Prunus persica var. nectarina) was performed using gas chromatography-isotope ratio mass spectrometry (GC-IRMS) in the combustion (C) and pyrolysis (P) mode.
Mohamed Alchihab et al.
Journal of industrial microbiology & biotechnology, 37(2), 167-172 (2009-11-11)
During the biotransformation of castor oil into gamma-decalactone, R. aurantiaca produced both the lactone form and its precursor (4-hydroxydecanoic acid). After six days of culture, a maximum yield of gamma-decalactone of 6.5 g/l was obtained. The parameters of gamma-decalactone adsorption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service