Skip to Content
Merck
All Photos(1)

Key Documents

W244503

Sigma-Aldrich

Ethyl myristate

≥98%, FCC, FG

Synonym(s):

Ethyl tetradecanoate, Myristic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOC2H5
CAS Number:
Molecular Weight:
256.42
FEMA Number:
2445
Beilstein:
1776382
EC Number:
Council of Europe no.:
385
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.104
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.436 (lit.)

bp

178-180 °C/12 mmHg (lit.)

mp

11-12 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

waxy; sweet

SMILES string

CCCCCCCCCCCCCC(=O)OCC

InChI

1S/C16H32O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16(17)18-4-2/h3-15H2,1-2H3

InChI key

MMKRHZKQPFCLLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

The substance W244503 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woo-kyoung Lee et al.
Journal of controlled release : official journal of the Controlled Release Society, 84(3), 115-123 (2002-12-07)
An oil-in-water solvent evaporation method was used to prepare cyclosporin A (CyA)-loaded particles varying in size (nanoparticles, 'small-sized' microparticles, 'large-sized' microparticles), polymer compositions [poly(D,L-lactide-co-glycolic acid) (PLGA) 50/50, PLGA 85/15, poly(D,L-lactic acid) (PLA)] and additive fatty acid ester (ethyl myristate; EM).
E Akaho et al.
Journal of pharmaceutical sciences, 70(11), 1225-1228 (1981-11-01)
Interactions of phenolic compounds 4-hexylresorcinol and 3,4-dimethylphenol with esters were studied using hexane-ester cosolvent systems by both phase solubility and partitioning methods. The data obtained by the phase solubility method were variable and could not be analyzed by any mathematical
V Auwärter et al.
Clinical chemistry, 47(12), 2114-2123 (2001-11-24)
Fatty acid ethyl esters (FAEEs) are products of nonoxidative ethanol metabolism. After incorporation in hair, they should be suitable long-term markers of alcohol abuse. Hair samples from 19 alcoholics in a treatment program, 10 fatalities with verified excessive alcohol consumption
Neonatal hair analysis as a biomarker for in utero alcohol exposure.
Julia Klein et al.
The New England journal of medicine, 347(25), 2086-2086 (2002-12-20)
Jennifer Peterson et al.
The Journal of pediatrics, 152(6), 788-792 (2008-05-22)
To determine the relationship between fatty acid ethyl esters (FAEE) in meconium and neurodevelopment in infants exposed to alcohol in utero at 6.5 months, 1 year, and 2 years of age. A secondary analysis of a prospective cohort of mothers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service