Skip to Content
Merck
All Photos(1)

Key Documents

W244910

Sigma-Aldrich

Ethyl octanoate

natural, ≥98%, FCC, FG

Synonym(s):

Ethyl caprylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOC2H5
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2449
Beilstein:
1754470
EC Number:
Council of Europe no.:
392c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.111
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor pressure

0.02 mmHg ( 25 °C)

Assay

≥98%

refractive index

n20/D 1.417 (lit.)

bp

206-208 °C (lit.)

mp

−48-−47 °C (lit.)

solubility

ethanol: soluble 1ml/4ml, clear, colorless (70% ethanol)

density

0.867 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana; fruity; floral; pineapple

SMILES string

CCCCCCCC(=O)OCC

InChI

1S/C10H20O2/c1-3-5-6-7-8-9-10(11)12-4-2/h3-9H2,1-2H3

InChI key

YYZUSRORWSJGET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl octanoate is an ethyl ester. It is an aroma active compound and is mainly responsible for the aroma of durian juices and wines. Its odor activity value (OAV) has been measured. It has been identified as an odor active compound present in the young monovarietal red wines prepared from various grape species.

Application

Ethyl octanoate is suitaible reagent used as standard for the measurement of flavor-active compounds by gas chromatography. It may be employed as acyl donor in acyl transfer reactions catalyzed by Candida antarctica lipase B in organic media and in lipase catalyzed acyl transfer reaction with 2-octanol as acyl acceptor.
Ethyl octanoate may be employed as acyl donor in the Candida antarctica lipase B catalyzed acyl transfer reactions. Reaction mechanism and kinetics of these reactions have been studied.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thioethyl-, vinyl-, ethyl octanoate esters and octanoic acid as acyl donors in lipase catalysed acyl transfer reactions.
Ohrner N, et al.
Biocatalysis and Biotransformation, 9(1-4), 105-114 (1994)
Quantitative determination of the odorants of young red wines from different grape varieties.
Ferreira V, et al.
Journal of the Science of Food and Agriculture, 80(11), 1659-1667 (2000)
JianCai Zhu et al.
Journal of agricultural and food chemistry, 63(7), 1939-1947 (2015-01-27)
The study evaluated the effects of five different strains (GRE, RC212, Lalvin D254, CGMCC2.4, and CGMCC2.23) of the yeast Saccharomyces cerevisiae on the aromatic characteristics of fermented durian musts. In this work, 38 and 43 compounds in durian juices and
Carrier-free, continuous primary beer fermentation.
Pires EJ, et al.
Journal of the Institute of Brewing, 120(4), 500-506 (2014)
M Martinelle et al.
Biochimica et biophysica acta, 1251(2), 191-197 (1995-09-06)
The acyl transfer reactions catalysed by Candida antartica lipase B in organic media followed a bi-bi ping-pong mechanism, with competitive substrate inhibition by the alcohols used as acyl acceptors. The effect of organic solvents on Vm and Km was investigated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service