Skip to Content
Merck
All Photos(1)

Key Documents

W242802

Sigma-Aldrich

Ethyl isobutyrate

≥98%, FCC, FG

Synonym(s):

Ethyl 2-methylpropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOOC2H5
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2428
Beilstein:
773846
EC Number:
Council of Europe no.:
288
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.413
NACRES:
NA.21
Organoleptic:
ethereal; fruity; sweet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

4.01 (vs air)

vapor pressure

40 mmHg ( 33.8 °C)

Assay

≥98%

refractive index

n20/D 1.387 (lit.)

bp

112-113 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

ethereal; fruity; sweet

SMILES string

CCOC(=O)C(C)C

InChI

1S/C6H12O2/c1-4-8-6(7)5(2)3/h5H,4H2,1-3H3

InChI key

WDAXFOBOLVPGLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl isobutyrate is a potent odorant in oranges, grapefruit, and Greek virgin olive oils.

Application


  • Recyclable polymeric pi-acid catalyst effective on Mannich-type reaction in water.: This study explores the use of a recyclable polymeric pi-acid catalyst, effective in facilitating Mannich-type reactions in aqueous environments, demonstrating a potential for enhancing green chemistry processes through water-based reactions (Masaki Y et al., 2006).


Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of key aroma compounds in hand-squeezed grapefruit juice (Citrus paradisi Macfayden) by quantitation and flavor reconstitution experiments.
Buettner A & Schieberle P.
Journal of Agricultural and Food Chemistry, 49(3), 1358-1363 (2001)
Evaluation and quantification of potent odorants of Greek virgin olive oils.
Blekas G & Guth H.
Developments in food science, 37, 419-427 (1995)
Evaluation of aroma differences between hand-squeezed juices from Valencia late and Navel oranges by quantitation of key odorants and flavor reconstitution experiments.
Buettner A & Schieberle P.
Journal of Agricultural and Food Chemistry, 49(5), 2387-2394 (2001)
Philippe Renault et al.
International journal of food microbiology, 207, 40-48 (2015-05-24)
The aim of this work was to study ester formation and the aromatic impact of Torulaspora delbrueckii when used in association with Saccharomyces cerevisiae during the alcoholic fermentation of must. In order to evaluate the influence of the inoculation procedure
Georgia Lytra et al.
Food chemistry, 194, 196-200 (2015-10-17)
Our study focused on variations in wine aroma perception and molecular composition during tasting over a period of 30min. In parallel, dynamic analytical and sensory methods were applied to study changes in the wines' molecular and aromatic evolution. Dynamic sensory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service