Skip to Content
Merck
All Photos(3)

Key Documents

W213810

Sigma-Aldrich

Benzyl benzoate

greener alternative

natural, ≥99%, FG

Synonym(s):

Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24
FEMA Number:
2138
Beilstein:
2049280
EC Number:
Council of Europe no.:
262
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.727
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 175.105

vapor pressure

1 mmHg ( 125 °C)

Assay

≥99%

form

liquid

autoignition temp.

896 °F

composition

contains IFRA and EU 1223/2009 restricted benzyl benzoate

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.568 (lit.)

bp

323-324 °C (lit.)

mp

17-20 °C (lit.)

density

1.118 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

benzyl benzoate

greener alternative category

Organoleptic

floral; fruity; balsamic; sweet

SMILES string

O=C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl benzoate is an aromatic ester that can be used as a flavoring agent.[1] It has been identified as one of the main volatile aroma component of cranberry{88], mango[2] and Egyptian Jasminum sambac flowers.[3]
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Toxicity and Repellency Efficacy of Benzyl Alcohol and Benzyl Benzoate as Eco-Friendly Choices to Control the Red Flour Beetle Tribolium castaneum.: This research investigates the toxicity and repellency of benzyl benzoate against the red flour beetle, highlighting its potential as an eco-friendly pest control agent (Aboelhadid et al., 2023).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 141-141 (2012)
Contribution of volatile compounds to mango (Mangifera indica L.) aroma
Pino JA & Mesa J
Flavour and Fragrance Journal, 21(2), 207-213 (2006)
Isolation and characterization of the volatile aroma compounds from the concrete headspace and the absolute of Jasminum sambac (L.) Ait.(Oleaceae) flowers grown in Egypt
Edris AE, et al
European Food Research and Technology, 226(3), 621-621 (2008)
Glen H MacDonald et al.
Hearing research, 243(1-2), 1-10 (2008-06-25)
The complex anatomy of the mammalian cochlea is most readily understood by representation in three-dimensions. However, the cochlea is often sectioned to minimize the effects of its anatomic complexity and optical properties on image acquisition by light microscopy. We have
Jing-Liang Li et al.
The journal of physical chemistry. B, 113(15), 5011-5015 (2009-03-25)
The formation of most organogels requires the compatibility of both the gelator and solvent. It is very desirable if the rheological properties of a gel can be manipulated to achieve the desired performance. In this paper, a novel organogel was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service