Skip to Content
Merck
All Photos(1)

Key Documents

T22403

Sigma-Aldrich

Tetrahydrothiophene 1-oxide

96%

Synonym(s):

Tetramethylene sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8OS
CAS Number:
Molecular Weight:
104.17
Beilstein:
105274
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.52 (lit.)

bp

235-237 °C (lit.)

density

1.158 g/mL at 25 °C (lit.)

SMILES string

O=S1CCCC1

InChI

1S/C4H8OS/c5-6-3-1-2-4-6/h1-4H2

InChI key

ISXOBTBCNRIIQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V K Chadha et al.
Journal of medicinal chemistry, 26(6), 916-922 (1983-06-01)
Sulfoxides and amides were tested as inhibitors of liver alcohol dehydrogenase and ethanol metabolism in rats. With both series of compounds, increasing the hydrophobicity resulted in better inhibition, and introduction of polar groups reduced inhibition. Of the cyclic sulfoxides, tetramethylene
Sangeun Yun et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(3), 204-211 (2019-12-06)
Lead halide perovskite is one of the attractive functional materials owing to its outstanding opto-electronic properties, which have been addressed in numerous studies. This study aims to clarify the link between the growth pattern and the charge carrier related properties
R Meganathan et al.
Journal of bacteriology, 169(6), 2862-2865 (1987-06-01)
Escherichia coli used tetrahydrothiophene 1-oxide (THTO) as an electron acceptor for anaerobic growth with glycerol as a carbon source; the THTO was reduced to tetrahydrothiophene. Cell extracts also reduced THTO to tetrahydrothiophene in the presence of a variety of electron
G G Balavoine et al.
Nitric oxide : biology and chemistry, 1(6), 507-521 (1997-01-01)
Kinetics and products of peroxynitrite anion O=NOO- reactions, catalyzed by water-soluble manganese and iron porphyrins, were studied under basic and neutral conditions. In the absence of organic substrates peroxynitrite decomposes catalytically to give nitrite and dioxygen as major products. Catalytic
R A Page et al.
The Biochemical journal, 278 ( Pt 3), 659-665 (1991-09-15)
We used titration with the inhibitors tetramethylene sulphoxide and isobutyramide to assess quantitatively the importance of alcohol dehydrogenase in regulation of ethanol oxidation in rat hepatocytes. In hepatocytes isolated from starved rats the apparent Flux Control Coefficient (calculated assuming a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service