Skip to Content
Merck
All Photos(2)

Key Documents

481858

Sigma-Aldrich

Methyl p-tolyl sulfoxide

97%

Synonym(s):

(±)-Methyl p-toluene sulfoxide, (±)-p-Tolyl methyl sulfoxide, 1-Methanesulfinyl-4-methylbenzene, 1-Methyl-4-(methylsulfinyl)benzene, 4-(Methylsulfinyl)toluene, 4-Methylphenyl methyl sulfoxide, 4-Toluene methyl sulfoxide, Methyl 4-methylphenyl sulfoxide, p-Methylphenyl methyl sulfoxide, p-Tolyl methyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4S(O)CH3
CAS Number:
Molecular Weight:
154.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

113-114 °C/2 mmHg (lit.)

mp

44-46 °C (lit.)

SMILES string

Cc1ccc(cc1)S(C)=O

InChI

1S/C8H10OS/c1-7-3-5-8(6-4-7)10(2)9/h3-6H,1-2H3

InChI key

FEVALTJSQBFLEU-UHFFFAOYSA-N

Related Categories

General description

Methyl p-tolyl sulfoxide (MTSO), an alkyl-substituted p-tolyl sulfoxide, is a Lewis base. It behaves as an activator of silicon tetrachloride that is employed in aza-Diels-Alder reaction.

Application

Methyl p-tolyl sulfoxide may be used as a catalyst in the preparation of homoallylic alcohols by the allylation of aldehydes with allyltrichlorosilane. It may be used as a ligand in the synthesis of molybdenum chlorocomplexes.

Caution

Low melting solid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

A New Approach to Di-and Tetrasubstituted 2,3-Dihydropyridin-4(1H)-ones through Aza-Diels-Alder Reaction Promoted by Silicon Tetrachloride.
Peduto A, et al.
Synthesis, 4, 0643-0649 (2009)
Asymmetric allylation of aldehydes with allyltrichlorosilane promoted by chiral sulfoxides.
Massa A, et al.
Tetrahedron Letters, 44(38), 7179-7181 (2003)
Luisa A Denkel et al.
PloS one, 6(11), e26974-e26974 (2011-11-11)
Production of reactive oxygen species represents a fundamental innate defense against microbes in a diversity of host organisms. Oxidative stress, amongst others, converts peptidyl and free methionine to a mixture of methionine-S- (Met-S-SO) and methionine-R-sulfoxides (Met-R-SO). To cope with such
Addition compounds of MoO2Cl2 with chiral sulfoxides. First molecular structures of dioxomolybdenum complexes bearing chiral non-racemic sulfoxide as ligand.
Pedrosa MR, et al.
Inorgorganica Chimica Acta, 363(13), 3158-3164 (2010)
Yuuka Masuyama et al.
Drug metabolism and pharmacokinetics, 35(3), 274-280 (2020-04-20)
Flavin containing monooxygenases (FMOs) represent one of the predominant types of phase I drug metabolizing enzymes (DMEs), and thus play an important role in the metabolism of xeno- and endobiotics for the generation of their corresponding oxides. These oxides often

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service