P7703
1,5-Pentanediol
96%
Synonym(s):
Pentamethylene glycol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO(CH2)5OH
CAS Number:
Molecular Weight:
104.15
Beilstein:
1560130
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
<0.01 mmHg ( 20 °C)
Assay
96%
autoignition temp.
635 °F
expl. lim.
13.2 %
refractive index
n20/D 1.450 (lit.)
bp
242 °C (lit.)
density
0.994 g/mL at 25 °C (lit.)
SMILES string
OCCCCCO
InChI
1S/C5H12O2/c6-4-2-1-3-5-7/h6-7H,1-5H2
InChI key
ALQSHHUCVQOPAS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
1,5-Pentanediol can be used:
- As a monomer for the synthesis of biocompatible polyesters, polycarbonates and polyurethanes.
- As an oxygen precursor for the formation of thin film of ZnO via atomic layer deposition.
- For the enantioselective convergent synthesis of (+)-spirolaxine methyl ether, ′helicobactericidal agent found in white rot fungi′.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
287.6 °F - closed cup
Flash Point(C)
142 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A formal total synthesis of anti-Helicobacter pylori agent (+)-spirolaxine methyl ether
Gadakh SK and Sudalai A
Tetrahedron Letters, 57(1), 25-28 (2016)
CH3ONa-initiated two-step-transesterification of DMC (dimethyl carbonate) and ?, ?-alkanediol for poly (alkylene carbonate)
Zhou R, et al.
Inorganic Chemistry Communications, 90, 82-85 (2018)
Biobased flexible aromatic polyester poly (1,5-pentylene terephthalate)(PPeT): Revisiting melt crystallization behaviors and thermo-mechanical properties
Lu J, et al.
European Polymer Journal, 110, 168-175 (2019)
Biodegradable shape memory polymers functionalized with anti-biofouling interpenetrating polymer networks
Dueramae I, et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 4(32), 5394-5404 (2016)
1, 5-Pentanediol as an Oxygen Precursor for Atomic Layer Deposition of Zinc Oxide Thin Films
Han SJ, et al.
Chemistry of Materials, 29(8), 3371-3374 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service