Skip to Content
Merck
All Photos(4)

Key Documents

M44451

Sigma-Aldrich

4,4′-Methylenebis(N,N-dimethylaniline)

97.5 - 102.5% purity, powder, crystals or chunks

Synonym(s):

N,N,N′,N′-Tetramethyl-4,4′-diaminodiphenylmethane, N,N,N′,N′-Tetramethyl-4,4′-methylenedianiline, Arnolds base, Michlers base

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H4N(CH3)2]2
CAS Number:
Molecular Weight:
254.37
Beilstein:
479956
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

4,4′-Methylenebis(N,N-dimethylaniline), 97.5% (GC)

Assay

97.5% (GC)

form

powder, crystals or chunks

technique(s)

titration: suitable

color

white to faint beige, to Tan

mp

88-89 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(Cc2ccc(cc2)N(C)C)cc1

InChI

1S/C17H22N2/c1-18(2)16-9-5-14(6-10-16)13-15-7-11-17(12-8-15)19(3)4/h5-12H,13H2,1-4H3

InChI key

JNRLEMMIVRBKJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Methylenebis(N,N-dimethylaniline) is an intermediate in dye manufacture and works as a reagent for the determination of lead. It is suitable for the qualitative assay of cyanogens.

Application

4,4′-Methylenebis(N,N-dimethylaniline) has been used to check the production of hydrogen cyanide by bacteria.
4,4′-Methylenebis(N,N-dimethylaniline) has been used as a component in the preparation of Feigl-Anger paper for visualizing HCN release. It may also be suitable for the preparation and characterization of inclusion complex with β-cyclodextrin.

Biochem/physiol Actions

4,4′-Methylenebis(N,N-dimethylaniline) is used in the adduct purification of precursors. It is used as an intermediate in the preparation of its hydrochloric salt.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is the ability of biocontrol fluorescent pseudomonads to produce the antifungal metabolite 2,4-diacetylphloroglucinol really synonymous with higher plant protection?
Rezzonico F
The New phytologist, 173, 861-861 (2007)
Eighth Annual Report on Carcinogens (1999)
A S Murthy
Toxicology letters, 6(6), 391-397 (1980-10-01)
Follicular neoplasms of the thyroid gland developed in 79 of 200 Fischer 344 rats of both sexes, fed either 0.0375% or 0.075% 4,4'-methylene-bis-(N,N-dimethyl)-benzenamine (MDBA) mixed with the diet. Hyperplastic changes in the follicular epithelium occurred in 12 rats fed the
4,4'-Methylenebis(N,N-dimethyl)benzenamine.
Report on carcinogens : carcinogen profiles, 11, III168-III168 (2004-01-01)
Cassava flour and starch: progress in research and development (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service