Skip to Content
Merck
All Photos(3)

Key Documents

515124

Sigma-Aldrich

N,N-Dimethylaniline

ReagentPlus®, 99%

Synonym(s):

N,N-Dimethylaniline, N,N-Dimethylphenylamine, DMA, Dimethylaniline, Dimethylphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(CH3)2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
application(s):
microbiology
bp:
193-194 °C (lit.)

Quality Level

product line

ReagentPlus®

Assay

99%

form

liquid

dilution

(for general lab use)

refractive index

n20/D 1.557 (lit.)

pH

7.4 (20 °C, 1.2 g/L)

bp

193-194 °C (lit.)

mp

1.5-2.5 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

application(s)

microbiology

functional group

amine

SMILES string

CN(C)c1ccccc1

InChI

1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

JLTDJTHDQAWBAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylaniline is a tertiary amine used in the synthesis of several triarylmethane dyes like malachite green. It is also used in the synthesis of a magnetic gram stain for the detection of bacteria.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A magnetic Gram stain for bacterial detection.
Budin, Ghyslain et al.
Angewandte Chemie (International Edition in English), 124(31), 7872-7875 (2012)
A synthetic study on the preparation of triarylmethanes.
Muthyala, Ramaiah et al.
Dyes and Pigments, 25(4), 303-324 (1994)
Triarylmethane Dyes for Artificial Repellent Cotton Fibers.
Montagut, Ana Maria et al.
Chemistry?A European Journal , 23(16), 3810-3814 (2017)
Marius Koch et al.
Journal of the American Chemical Society, 134(8), 3729-3736 (2012-01-31)
The fluorescence quenching of 3-cyanoperylene upon electron transfer from N,N-dimethylaniline in three room-temperature ionic liquids (RTILs) and in binary solvent mixtures of identical viscosity has been investigated using steady-state and time-resolved fluorescence spectroscopy. This study was stimulated by previous reports
Yong Wang et al.
The journal of physical chemistry. B, 114(8), 2964-2970 (2010-02-12)
This paper addresses the experimentally observed mechanistic differences between the cytochrome P450-catalyzed N-demethylation of substituted N,N-dimethylanilines (DMA) and of N,N-dimethylbenzamides (DMBA). The two reactions of these substrates are initiated by C-H activation of the methyl groups on the nitrogen. Thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service