Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium
Synonym(s):
Lithium
Linear Formula:
Li
CAS Number:
7439-93-2
Molecular Weight:
6.94
Products
Brand
Sigma-Aldrich (12)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Diameter (mm)
Element
Quality Segment
Application
Reagent Type
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
499811
Lithium,
granular, 99% trace metals basis
Expand
View Pricing
265985
Lithium,
ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Expand
View Pricing
GF47159826
Lithium,
foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%
Expand
View Pricing
444456
Lithium,
granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Expand
View Pricing
320080
Lithium,
ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Expand
View Pricing
265969
Lithium,
rod, diam. 12.7 mm, 99.9% trace metals basis
Expand
View Pricing
266000
Lithium,
ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Expand
View Pricing
278327
Lithium,
wire, diam. 3.2 mm, in mineral oil, ≥98%
Expand
View Pricing
265993
Lithium,
ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Expand
View Pricing
220914
Lithium,
wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Expand
View Pricing
8.05660
Lithium,
(sticks) (protective liquid: paraffin oil) for synthesis
Expand
View Pricing
GF11597913
Lithium,
foil, thickness 0.6 mm, size 25 × 300 mm, purity 99.9%
Expand
View Pricing
GF62158596
Lithium,
foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%
Expand
View Pricing
8.18044
Lithium,
(granular) (protective medium: argon) for synthesis
Expand
View Pricing
GF45554799
Lithium,
rod, 12.7 mm diameter, length 165 mm, purity 99%
Expand
View Pricing
GF40266682
Lithium,
foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%
Expand
View Pricing