Skip to Content
Merck
All Photos(2)

Key Documents

GF27295724

Cerium

foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%

Synonym(s):

Cerium, CE000210

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ce
CAS Number:
Molecular Weight:
140.12
MDL number:
UNSPSC Code:
12141601
PubChem Substance ID:
NACRES:
NA.23

Assay

99.90%

form

foil

manufacturer/tradename

Goodfellow 272-957-24

resistivity

73 μΩ-cm, 20°C

size × thickness

50x50 mm × 0.1 mm

bp

3443 °C (lit.)

mp

795 °C (lit.)

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ce]

InChI

1S/Ce

InChI key

GWXLDORMOJMVQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Jakupec et al.
Reviews of physiology, biochemistry and pharmacology, 153, 101-111 (2005-01-28)
Cerium is a member of the lanthanide series or rare earth elements which exert diverse biological effects mainly by their resemblance to calcium. This similarity, which is particularly characteristic for the lighter members of the lanthanide series, enables these elements
J P Garner et al.
Burns : journal of the International Society for Burn Injuries, 31(5), 539-547 (2005-06-16)
The introduction of early excision of the burn eschar has contributed to a reduction in burn-related mortality but is not appropriate in all circumstances. Cerium nitrate has been used since 1976, usually in combination with silver sulphadiazine, to improve outcome
Ali Bumajdad et al.
Advances in colloid and interface science, 147-148, 56-66 (2008-11-26)
This review concerns recent research on the synthesis of cerium oxide (also known as ceria, CeO(2)) in colloidal dispersions media for obtaining high surface area catalyst materials. Nanoparticles as small as 5 nm and surface area as high as 250
J W McDonald et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 8(8), 859-865 (1995-10-01)
Rare earth pneumoconiosis is an uncommonly reported disease caused by the inhalation of dust containing lanthanides, also known as rare earth metals, which are common industrial materials. The pathologic manifestations and natural history of this disorder are incompletely understood. We
Soumen Das et al.
Nanomedicine (London, England), 8(9), 1483-1508 (2013-08-31)
Promising results have been obtained using cerium (Ce) oxide nanoparticles (CNPs) as antioxidants in biological systems. CNPs have unique regenerative properties owing to their low reduction potential and the coexistence of both Ce(3+)/Ce(4+) on their surfaces. Defects in the crystal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service