460877
Neodymium
powder, −40 mesh, ≥99% trace rare earth metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Nd
CAS Number:
Molecular Weight:
144.24
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
≥99% trace rare earth metals basis
form
powder
reaction suitability
reagent type: catalyst
core: neodymium
resistivity
64.0 μΩ-cm, 20°C
particle size
−40 mesh
bp
3074 °C (lit.)
mp
1021 °C (lit.)
density
7.003 g/mL at 25 °C (lit.)
SMILES string
[Nd]
InChI
1S/Nd
InChI key
QEFYFXOXNSNQGX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Sol. 2 - Self-heat. 1 - Skin Irrit. 2
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ionut Mihalcea et al.
Inorganic chemistry, 51(18), 9610-9618 (2012-08-31)
Three series of mixed uranyl-lanthanide (Ce or Nd) carboxylate coordination polymers have been successfully synthesized by means of a hydrothermal route using either conventional or microwave heating methods. These compounds have been prepared from mixtures of uranyl nitrate, lanthanide nitrate
Angie Phelan et al.
Australian orthodontic journal, 28(2), 159-169 (2013-01-12)
The demand for clear sequential aligner therapy has increased dramatically in recent years. An improved system utilising small neodymium-iron-boron (NdFeB) magnetic attachments has been proposed to enhance appliance capabilities. The aim of the investigation was to analyse the force system
Haohai Yu et al.
Scientific reports, 3, 1085-1085 (2013-01-22)
Two-dimensional (2D) visible crystal-structure patterns analogous to the quantum harmonic oscillator (QHO) have been experimentally observed in the near- and far-fields of a self-frequency-doubling (SFD) microchip laser. Different with the fundamental modes, the localization of the SFD light is changed
Johanna Laakso et al.
Inorganic chemistry, 51(19), 10366-10374 (2012-09-18)
Lanthanide (Yb(3+), Nd(3+)) complexes equipped with red-absorbing hydroporphyrin (chlorin) antennae were synthesized and characterized. The syntheses are scalable, highly modular, and enable the introduction of different chlorins functionalized with a single reactive group (COOH or NH(2)). Absorption maxima were dependent
Wei-Bin Cui et al.
Advanced materials (Deerfield Beach, Fla.), 24(48), 6530-6535 (2012-10-31)
Anisotropic Nd(2) Fe(14) B/Fe(67) Co(33) nanocomposite thin films are successfully fabricated. The multilayer composite films comprise Nd-rich shell-enveloped Nd(2) Fe(14) B grains and a Fe(67) Co(33) phase. The strong (001) texture of the Nd(2) Fe(14) B grains and the presence
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service