Skip to Content
Merck
All Photos(2)

Key Documents

GF15174390

Gadolinium

rod, 50mm, diameter 6.35mm, cast, 99%

Synonym(s):

Gadolinium, GD007910

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd
CAS Number:
Molecular Weight:
157.25
MDL number:
UNSPSC Code:
12141605
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

rod

manufacturer/tradename

Goodfellow 151-743-90

resistivity

126 μΩ-cm, 20°C

L × diam.

50 mm × 6.35 mm

bp

3273 °C (lit.)

mp

1313 °C (lit.)

density

7.886 g/mL at 25 °C (lit.)

SMILES string

[Gd]

InChI

1S/Gd

InChI key

UIWYJDYFSGRHKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongjun Liu et al.
Biomaterials, 33(21), 5363-5375 (2012-04-24)
Theranostic magnetic resonance imaging (MRI) is now receiving a growing interest in imaging-guided drug delivery, monitoring the treatment and personalized administration etc. Theranostic agents are essential for the usage of theranostic MRI. Among different kinds of theranostic agents, gadolinium loaded
Andrew J Swift et al.
JACC. Cardiovascular imaging, 7(12), 1209-1217 (2014-12-17)
The goal of this study was to determine the prognostic value of late gadolinium enhancement (LGE) cardiac magnetic resonance (CMR) features in patients with pulmonary hypertension. The prognostic significance of LGE in the clinical assessment of patients with pulmonary hypertension
Wendy Oakden et al.
NMR in biomedicine, 28(8), 925-936 (2015-06-09)
Quantitative T2 (qT2), diffusion tensor imaging (DTI), and histology were used to investigate a cervical model of spinal cord injury (SCI) in the rat. While quantitative MRI can significantly increase the specificity in the presence of pathology, it must be
R Jakubovic et al.
Clinical oncology (Royal College of Radiologists (Great Britain)), 26(11), 704-712 (2014-07-16)
To investigate whether early relative cerebral blood volume (rCBV), relative cerebral blood flow (rCBF) and permeability (Ktrans(2)) measurements may serve as magnetic resonance imaging (MRI) biomarkers of radiation response or progression for brain metastases. Seventy brain metastases in 44 patients
Brigit den Adel et al.
Atherosclerosis, 225(2), 274-280 (2012-08-14)
MRI using targeted contrast agents (CA) has emerged as a promising technique to study atherothrombotic disease in vivo. Particularly, the use of targeted Gd and lipid-based nanoparticles has enabled detailed in vivo imaging of various molecular markers of atherosclerotic plaque

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service