Skip to Content
Merck
All Photos(1)

Key Documents

D42607

Sigma-Aldrich

1,8-Dibromooctane

98%

Synonym(s):

Octamethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)8Br
CAS Number:
Molecular Weight:
272.02
Beilstein:
1698114
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

270-272 °C (lit.)

mp

12-16 °C (lit.)

density

1.477 g/mL at 25 °C (lit.)

SMILES string

BrCCCCCCCCBr

InChI

1S/C8H16Br2/c9-7-5-3-1-2-4-6-8-10/h1-8H2

InChI key

DKEGCUDAFWNSSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,8-Dibromooctaneis primarily used in organic synthesis as a building block for the preparationof various chemical intermediates.It is used as an alkylating reagent in aone-pot synthesis for the production of paraffin-like oligomers and it is usedto enhance crystallization and film formation of perovskite.

Application


  • Efficient manual Fmoc solid-phase synthesis of the N-terminal segment of surfactant protein B (SP-B(1-25)).: This study highlights the application of Fmoc-Gly-Wang resin in the synthesis of peptide segments, specifically demonstrating its utility in constructing the N-terminal segment of surfactant protein B (SP-B). The research underscores the resin′s effectiveness in peptide synthesis, emphasizing its role in generating biochemically significant peptides (Ramli et al., 2009).


Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From N-vinylpyrrolidone anions to modified paraffin-like oligomers via double alkylation with 1, 8-dibromooctane: access to covalent networks and oligomeric amines for dye attachment
Obels, Daniela and Lievenbr{"u}ck, et, al.
Beilstein Journal of Organic Chemistry, 12, 1395-1400 (2016)
Efficiency enhancement in planar CH3NH3PbI3- xClx perovskite solar cells by processing with bidentate halogenated additives
Fu, Guangsheng and Hou, et al.
Solar Energy Materials and Solar Cells, 165, 36-44 (2017)
Enrico Caruso et al.
Bioorganic & medicinal chemistry, 28(21), 115737-115737 (2020-10-17)
A new class of compounds based on the 4,4-difluoro-4-bora-3a,4a-diaza-s-indacene core, known as BODIPYs, has attracted significant attention as photosensitizers suitable for application in photodynamic therapy (PDT), which is a minimally invasive procedure to treat cancer. In PDT the combination of
Kenji Tajima et al.
Biomacromolecules, 21(2), 581-588 (2019-11-22)
Nanofibrillated bacterial cellulose (NFBC) is produced by culturing a cellulose-producing bacterium under agitated aerobic conditions in a carboxymethylcellulose (CMC)-supplemented medium. Detailed structural analyses revealed that NFBC fiber widths varied with the degree of substitution of the CMC used, and zeta
Enrico Caruso et al.
Journal of photochemistry and photobiology. B, Biology, 195, 39-50 (2019-05-11)
Photodynamic therapy (PDT) of cancer uses photosensitizers (PS), a light source and oxygen to generate high levels of reactive oxygen species (ROS), that exert a cytotoxic action on tumor cells. Recently, it has been shown that mixed non-symmetrical diaryl porphyrins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service