All Photos(1)
About This Item
Linear Formula:
I(CF2)8I
CAS Number:
Molecular Weight:
653.87
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
73-77 °C (lit.)
functional group
fluoro
iodo
storage temp.
2-8°C
SMILES string
FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
InChI
1S/C8F16I2/c9-1(10,3(13,14)5(17,18)7(21,22)25)2(11,12)4(15,16)6(19,20)8(23,24)26
InChI key
SRDQTCUHAMDAMG-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Liang Lu et al.
Journal of applied toxicology : JAT, 39(7), 945-954 (2019-03-06)
Fluorinated diiodine alkanes (FDIAs), important industrial intermediates in the synthesis of various perfluorinated compounds, which are distributed widely in wildlife and humans. Recent studies showed that FDIAs had in vitro estrogenic effects. However, to date, little information is available regarding
Wenting Song et al.
Talanta, 175, 413-420 (2017-08-27)
Contaminants of emerging concern are continuously increasing, which makes it important to develop high throughput screening techniques for the evaluation of their potential biological effects, especially endocrine disrupting effects, which would directly influence the population dynamics in environment. A novel
Chang Wang et al.
Environmental health perspectives, 120(1), 119-125 (2011-10-13)
Polyfluorinated iodine alkanes (PFIs) are important intermediates in the synthesis of organic fluoride products. Recently, PFIs have been detected in fluoropolymers as residual raw materials, as well as in the ambient environment. High production volumes and potential environmental releases of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service