D146706
9,10-Dimethylanthracene
99%
Synonym(s):
9,10-Dimethylanthracene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H14
CAS Number:
Molecular Weight:
206.28
Beilstein:
1909028
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
crystals
mp
182-184 °C (lit.)
SMILES string
Cc1c2ccccc2c(C)c3ccccc13
InChI
1S/C16H14/c1-11-13-7-3-5-9-15(13)12(2)16-10-6-4-8-14(11)16/h3-10H,1-2H3
InChI key
JTGMTYWYUZDRBK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shimshon Ben Dror et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 8(3), 354-361 (2009-03-04)
In this work we investigate the localization and photophysical properties of twelve synthetically derived chlorins in artificial membranes, with the goal of designing more effective photosensitizers for photodynamic therapy (PDT). The studied chlorins incorporate substituents of varying lipophilicity at the
Sankarasharma Devipriya et al.
Chemico-biological interactions, 162(2), 106-113 (2006-07-19)
Administration of quercetin, a common polyphenolic component of many vascular and edible plants including vegetables, fruits and tea significantly reduced the tumor volume in rats induced for mammary carcinoma using dimethyl benz (a) anthracene (DMBA). Dose response was assessed, by
A smart polysaccharide/drug conjugate for photodynamic therapy.
So Young Park et al.
Angewandte Chemie (International ed. in English), 50(7), 1644-1647 (2011-02-11)
Alan Sholto et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 7(3), 344-351 (2008-04-05)
Six amphiphilic silicon phthalocyanines (SiPc's) axially linked to a dimethylated amino alkyl group of varying length have been examined for their potential suitability as photosensitizers for photodynamic therapy (PDT). This group of molecules was chosen because the length of the
R K Saini et al.
Drug and chemical toxicology, 31(4), 459-471 (2008-10-14)
A single intratracheal instillation of 9,10-dimethyl benz(a)anthracene (DMBA) at 3 different doses of 5, 10, and 20 mg/kg body weight to Balb/c mice for 12 weeks had caused a significant incidence of pulmonary tumors along with inflammatory changes. The number
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service